Skip to Content
Merck
All Photos(1)

Key Documents

204609

Sigma-Aldrich

Thallium(I) nitrate

99.999% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TlNO3
CAS Number:
Molecular Weight:
266.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.999% trace metals basis

form

solid

reaction suitability

core: thallium

mp

206 °C (lit.)

SMILES string

[Tl+].[O-][N+]([O-])=O

InChI

1S/NO3.Tl/c2-1(3)4;/q-1;+1

InChI key

FYWSTUCDSVYLPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Thallium(I) nitrate: Primarily used in the synthesis of other thallium compounds due to its reactivity and solubility. It finds applications in the production of special glasses where thallium improves the refractive index and density of the glass. Caution is advised in its use due to its high toxicity and potential environmental impact (Sigma-Aldrich, CAS 10102-45-1).

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - Ox. Sol. 3 - STOT RE 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cecilia Eliana Hanzel et al.
Toxicology and applied pharmacology, 236(1), 59-70 (2009-04-18)
Thallium (Tl) is a highly toxic metal though yet its mechanisms are poorly understood. Previously, we demonstrated that rat pheochromocytoma (PC12) cells exposure to thallous (Tl(I)) or thallic (Tl(III)) cations leads to mitochondrial damage and reduced cell viability. In the
Vânia M T Carneiro et al.
The Journal of organic chemistry, 75(9), 2877-2882 (2010-04-10)
A diastereoselective route to (+)-bakkenolide A is presented from the readily available optically active Wieland-Miescher ketone. This novel synthesis of this sesquiterpene lactone features the following as key stereoselective transformations: (i) the ring contraction reaction of a octalone mediated by
María T L Pino et al.
Journal of applied toxicology : JAT, 35(8), 952-969 (2014-12-24)
The effects of thallium [Tl(I) and Tl(III)] on the PC12 cell cycle were evaluated without (EGF(-)) or with (EGF(+)) media supplementation with epidermal growth factor (EGF). The following markers of cell-cycle phases were analyzed: cyclin D1 (G1 ); E2F-1, cyclin
Tl+ increases the permeability of the inner membranes of rat liver mitochondria for monovalent cations.
S M Korotkov et al.
Doklady. Biochemistry and biophysics, 378, 145-149 (2001-11-20)
I A Skul'skiĭ et al.
Zhurnal evoliutsionnoi biokhimii i fiziologii, 24(6), 817-821 (1988-11-01)
Studies have been made of the effect of inhibitors of energy metabolism on maintenance of Na and K concentrations, as well as on accumulation of Rb (as an analogue of K) in single neurones of the mollusc P. corneus. Concentrations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service