303259
1-Chloro-3-methyl-2-butene
95%
Synonym(s):
γ,γ-Dimethylallyl chloride, 1,1-Dimethyl-3-chloro-1-propene, 1-Chloro-3-methyl-2-butene, 2-Methyl-4-chloro-2-butene, 3,3-Dimethylallyl chloride, 3-Methyl-2-butenyl chloride, 3-Methylcrotyl chloride, 4-Chloro-2-methyl-2-butene
About This Item
Recommended Products
Assay
95%
form
liquid
refractive index
n20/D 1.4488 (lit.)
bp
58-59 °C/120 mmHg (lit.)
density
0.928 g/mL at 25 °C (lit.)
functional group
alkyl halide
chloro
storage temp.
2-8°C
SMILES string
C\C(C)=C\CCl
InChI
1S/C5H9Cl/c1-5(2)3-4-6/h3H,4H2,1-2H3
InChI key
JKXQKGNGJVZKFA-UHFFFAOYSA-N
General description
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
55.4 °F - closed cup
Flash Point(C)
13 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service