Skip to Content
Merck
All Photos(1)

Documents

302791

Sigma-Aldrich

Geranyl chloride

95%

Synonym(s):

trans-1-Chloro-3,7-dimethyl-2,6-octadiene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCH2Cl
CAS Number:
Molecular Weight:
172.69
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.4808 (lit.)

bp

102-104 °C/12 mmHg (lit.)

density

0.931 g/mL at 25 °C (lit.)

SMILES string

C\C(C)=C\CC\C(C)=C\CCl

InChI

1S/C10H17Cl/c1-9(2)5-4-6-10(3)7-8-11/h5,7H,4,6,8H2,1-3H3/b10-7+

InChI key

WLAUCMCTKPXDIY-JXMROGBWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Geranyl chloride undergoes palladium-catalyzed cross-coupling reaction with aryl and alkenylgold(I) phosphanes to afford the α-substitution product.

Application

Geranyl chloride was used as starting reagent in the synthesis of:
  • C25 compound moenocinol
  • 2-methyl-6-geranylphenol
  • manoalide and seco-manoalide analogs

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A synthesis of moenocinol from isoprenoid precursors.
Bottger D and Welzel P.
Tetrahedron Letters, 24(47), 5201-5204 (1983)
Toward elucidation of the inhibition mechanism of phospholipase A< sub> 2</sub> by manoalide: selectively modified amino acid residues by manoalide analogues.
Katsumura S, et al.
Bioorganic & Medicinal Chemistry Letters, 2(10), 1267-1268 (1992)
Preparation and oxidative polymerization of 2-methyl-6-geranylphenol.
Hyun SH, et al.
Polymer Bull., 18(4), 283-286 (1987)
Miguel Peña-López et al.
Organic & biomolecular chemistry, 10(8), 1686-1694 (2012-01-24)
Aryl and alkenylgold(I) phosphanes react regioselectively with allylic electrophiles such as cinnamyl and geranyl halides (bromide, chloride and acetates) under palladium catalysis in THF at 80 °C to afford the α-substitution product with moderate to high yields. When the reaction
Minju Lee et al.
Scientific reports, 8(1), 13945-13945 (2018-09-19)
We report a strategy for the human-like smelling of a rose scent utilizing olfactory receptor nanodisc (ND)-based bioelectronic nose devices. In this strategy, a floating electrode (FE)-based carbon nanotube (CNT) field effect transistor (FET) was functionalized with human olfactory receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service