Skip to Content
Merck
All Photos(2)

Documents

301841

Sigma-Aldrich

1,7-Phenanthroline

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8N2
CAS Number:
Molecular Weight:
180.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

79-81 °C (lit.)

SMILES string

c1cnc2c(c1)ccc3ncccc23

InChI

1S/C12H8N2/c1-3-9-5-6-11-10(4-2-7-13-11)12(9)14-8-1/h1-8H

InChI key

OZKOMUDCMCEDTM-UHFFFAOYSA-N

General description

1,7-Phenanthroline has inhibitory effect on germination and growth of plants. 1,7-Phenanthroline is an aza-analog of phenanthrene, was found to be mutagenic in the Ames test using Salmonella typhimurium TA100 in the presence of a rat liver S9 fraction. Supramolecular assemblies of 1,2,4,5-benzenetetracarboxylic acid with 1,7-phenanthroline (aza donor molecule) were investigated.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kapildev K Arora et al.
The Journal of organic chemistry, 68(24), 9177-9185 (2003-11-25)
Supramolecular assemblies of 1,2,4,5-benzenetetracarboxylic acid, 1, with aza donor molecules such as 1,10-phenanthroline, 2, 1,7-phenanthroline, 3, phenazine, 4, 4-(N,N-dimethylamino)pyridine, 5, 1,2-bis(4-pyridyl)ethene, 6, and 1,2-bis(4-pyridyl)ethane, 7, have been synthesized and characterized by single-crystal X-ray diffraction methods. All the complexes crystallize in
Wei Xiao et al.
Environmental science & technology, 52(1), 114-123 (2017-12-06)
Iron (oxyhydr)oxides are widespread in natural and engineered systems, potent adsorbents of contaminants and a source of energy for iron-reducing bacteria. Microbial reduction of iron (oxyhydr)oxides results in the formation of Fe(II) which can induce the transformation of these iron
Martin Saurer et al.
Science (New York, N.Y.), 365(6458), 1144-1149 (2019-09-14)
Mitochondrial ribosomes (mitoribosomes) are large ribonucleoprotein complexes that synthesize proteins encoded by the mitochondrial genome. An extensive cellular machinery responsible for ribosome assembly has been described only for eukaryotic cytosolic ribosomes. Here we report that the assembly of the small
Katsuya Yamada et al.
Mutation research, 559(1-2), 83-95 (2004-04-07)
Phenanthrene, a simplest angular polycyclic aromatic hydrocarbon with a bay-region in its molecule, is reported to be non-mutagenic, although most angular (non-linear) polycyclic aromatic hydrocarbons, such as benzo[a]pyrene and chrysene, are known to show genotoxicity after metabolic transformation into a
Hyung Min Kim et al.
Molecules (Basel, Switzerland), 25(4) (2020-02-16)
Panax ginseng (P. ginseng) is the most widely consumed herbal plant in Asia and is well-known for its various pharmacological properties. Many studies have been devoted to this natural product. However, polysaccharide's components of ginseng and their biological effects have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service