171778
2-Aminobenzimidazole
97%
Synonym(s):
2-Benzimidazolamine
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C7H7N3
CAS Number:
Molecular Weight:
133.15
Beilstein:
116525
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
32151902
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
mp
226-230 °C (lit.)
SMILES string
Nc1nc2ccccc2[nH]1
InChI
1S/C7H7N3/c8-7-9-5-3-1-2-4-6(5)10-7/h1-4H,(H3,8,9,10)
InChI key
JWYUFVNJZUSCSM-UHFFFAOYSA-N
Gene Information
human ... PLAU(5328)
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
2-Aminobenzimidazole was used in the hydrolysis of a choline carbonate. It was also used in the synthesis of imidazo[1,2-a]benzimidazoles.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Bartolomé Soberats et al.
Organic letters, 16(3), 840-843 (2014-01-15)
The hydrolysis of a choline carbonate through a metal-free, enzyme-like mechanism has been achieved using a 2-aminobenzimidazole-based deep cavitand as catalyst. The supramolecular catalysis involves three steps: host-guest binding, carbamoylation and enzyme-like hydrolysis. Interestingly the rate-determining step proceeds through a
Ya-Shan Hsiao et al.
ACS combinatorial science, 15(10), 551-555 (2013-09-11)
A one-pot, two-step synthesis of imidazo[1,2-a]benzimidazoles has been achieved by a three-component reaction of 2-aminobenzimidazoles with an aromatic aldehyde and an isocyanide. The reaction involving condensation of 2-aminobenzimidazole with an aldehyde is run under microwave activation to generate an imine
S Sudha et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 84(1), 184-195 (2011-10-11)
In the present work, we reported a combined experimental and theoretical study on molecular structure, vibrational spectra and HOMO-LUMO analysis of 2-aminobenzimidazole (2-ABD). The FTIR (400-4000 cm(-1)) and FT-Raman spectra (50-3500 cm(-1)) of 2-ABD were recorded. The molecular geometry, harmonic
Zinc fingered: new compounds that thwart gram-positive biofilm formation by sequestering zinc.
Dinty J Musk
Chembiochem : a European journal of chemical biology, 11(6), 758-760 (2010-03-02)
Timothy Coon et al.
Bioorganic & medicinal chemistry letters, 19(15), 4380-4384 (2009-06-26)
The benzimidazole core of the selective non-brain-penetrating H(1)-antihistamine mizolastine was used to identify a series of brain-penetrating H(1)-antihistamines for the potential treatment of insomnia. Using cassette PK studies, brain-penetrating H(1)-antihistamines were identified and in vivo efficacy was demonstrated in a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service