Skip to Content
Merck
All Photos(1)

Key Documents

298301

Sigma-Aldrich

Pentaamminechlorocobalt(III) chloride

98%

Synonym(s):

Cobalt(III) chloropentammine chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Co(NH3)5Cl]Cl2
CAS Number:
Molecular Weight:
250.44
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Assay

98%

form

powder

SMILES string

N.N.N.N.N.Cl[Co](Cl)Cl

InChI

1S/3ClH.Co.5H3N/h3*1H;;5*1H3/q;;;+3;;;;;/p-3

InChI key

NSYALVBBDKTCLE-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

A nanocomposite of nanophotoadduct of pentaamminechlorocobalt(III) chloride with hexamine and polythiophene (PTh)may be used in high temperatures electrical appliances. It may be used along with potassium tris(oxalato)chromate(III) to synthesize heterometallic compound, [CoCl(NH3 )5 ] [KCr(C2 O4 )3 ]•0.5H2O.This heterometallic compound has several potential applications such as microporous hybrid material with organic π-donors.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Carc. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, characterization, electrical and thermal properties of nanocomposite of polythiophene with nanophotoadduct: a potent composite for electronic use.
Najar MH & Majid K
Journal of Materials Science: Materials in Electronics, 24.11, 4332-4339 (2013)
Unusual potassium-oxalate coordination in the two-dimensional trimetallic [CoCl(NH3 )5 ] [KCr(C2 O4 )3 ]?0.5H2O complex.
Wrzeszcz G, et al.
open chemistry, 12.6, 652-658 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service