Skip to Content
Merck
All Photos(1)

Documents

260983

Sigma-Aldrich

Tris(ethylenediamine)cobalt(III) chloride dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2NCH2CH2NH2)3CoCl3·2H2O
CAS Number:
Molecular Weight:
381.62
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

reaction suitability

core: cobalt
reagent type: catalyst

mp

275 °C (dec.) (lit.)

SMILES string

O.O.NCCN.NCCN.NCCN.Cl[Co](Cl)Cl

InChI

1S/3C2H8N2.3ClH.Co.2H2O/c3*3-1-2-4;;;;;;/h3*1-4H2;3*1H;;2*1H2/q;;;;;;+3;;/p-3

InChI key

ALLUKJGRPOLBEF-UHFFFAOYSA-K

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zari Hooshyar et al.
Luminescence : the journal of biological and chemical luminescence, 30(5), 538-548 (2014-10-15)
Diamine-sarcophagine (DiAmsar) binding to human serum albumin (HSA) and bovine serum albumin (BSA) was investigated under simulative physiological conditions. Fluorescence spectra in combination with Fourier transform infrared (FT-IR), UV-visible (UV-vis) spectroscopy, cyclic voltammetry (CV), and molecular docking method were used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service