295183
Cyclopropane
≥99%
Synonym(s):
Trimethylene (6CI)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C3H6
CAS Number:
Molecular Weight:
42.08
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor density
1.45 (vs air)
Assay
≥99%
autoignition temp.
928 °F
expl. lim.
10.4 %
bp
−33 °C (lit.)
mp
−128 °C (lit.)
SMILES string
C1CC1
InChI
1S/C3H6/c1-2-3-1/h1-3H2
InChI key
LVZWSLJZHVFIQJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Cyclopropane has been used as the pseudo-π unsaturated hydrocarbon prototype to study the rotational spectrum of its complex formed with chlorine monofluoride and the similarity of this complex with isotopomers.
Packaging
Supplied in a Sure/Pac™ cylinder and has a brass needle valve with a male 1/4" NPTF outlet thread installed. Before using the cylinder, ensure that the valve is closed, then remove the galvanized steel hex cap that seals the outlet valve.
Compatible with the following:
Compatible with the following:
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC
also commonly purchased with this product
Product No.
Description
Pricing
hose barb
Product No.
Description
Pricing
recommended
Product No.
Description
Pricing
regulator
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Gas 1 - Press. Gas Liquefied gas
Storage Class Code
2A - Gases
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Ya Zhang et al.
Frontiers in microbiology, 8, 2036-2036 (2017-11-04)
The occurrence of pathogenic bacteria in drinking water distribution systems (DWDSs) is a major health concern, and our current understanding is mostly related to pathogenic species such as
Joshua S Alford et al.
Organic letters, 14(23), 6020-6023 (2012-11-17)
Warming of 4-phthalimido-N-mesyl-1,2,3-triazole in the presence of alkenes followed by silica gel induced hydrolysis results in a highly diastereoselective and catalyst-free entry to N-phthalimidocyclopropanecarboxaldehydes.
Lorenzo Cianni et al.
PLoS neglected tropical diseases, 14(3), e0007755-e0007755 (2020-03-13)
The cysteine protease cruzipain is considered to be a validated target for therapeutic intervention in the treatment of Chagas disease. A series of 26 new compounds were designed, synthesized, and tested against the recombinant cruzain (Cz) to map its S1/S1´
Thomas Schalck et al.
Microorganisms, 9(2) (2021-02-04)
Fuels and polymer precursors are widely used in daily life and in many industrial processes. Although these compounds are mainly derived from petrol, bacteria and yeast can produce them in an environment-friendly way. However, these molecules exhibit toxic solvent properties
Zhiqiang Wen et al.
Microbial biotechnology, 13(2), 410-422 (2019-08-27)
Butanol is an important bulk chemical, as well as a promising renewable gasoline substitute, that is commonly produced by solventogenic Clostridia. The main cost of cellulosic butanol fermentation is caused by cellulases that are required to saccharify lignocellulose, since solventogenic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service