All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H7N3O2
CAS Number:
Molecular Weight:
153.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
solid
mp
223-225 °C (lit.)
functional group
nitro
SMILES string
Cc1cc(N)ncc1[N+]([O-])=O
InChI
1S/C6H7N3O2/c1-4-2-6(7)8-3-5(4)9(10)11/h2-3H,1H3,(H2,7,8)
InChI key
GRBBNZYMXKTQAI-UHFFFAOYSA-N
Related Categories
General description
The crystal structure of 2-amino-4-methyl-5-nitropyridine was elucidated.
Application
2-Amino-4-methyl-5-nitropyridine was used in the preparation of matrix mixture required to study new technical developments for the direct tissue analysis of peptides.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
R Lemaire et al.
Analytical chemistry, 78(3), 809-819 (2006-02-02)
Direct analysis of tissue by MALDI-MS allows the acquisition of its biomolecular profile while maintaining the integrity of the tissue, giving cellular localization, and avoiding tedious extraction and purification steps. However, direct tissue analysis generally leads to some extent to
I Bryndal et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 96, 952-962 (2012-09-04)
The crystal structures of 2-amino-4-methyl-3-nitropyridine (I), 2-amino-4-methyl-3,5-dinitropyridine (II) and 2-amino-4-methyl-5-nitropyridine (III) have been determined. The compounds crystallize in the monoclinic P2(1)/n, triclinic P-1 and monoclinic C2/c space groups, respectively. These structures are stabilized by a combination of N-H···N and N-H···O
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service