Skip to Content
Merck
All Photos(2)

Key Documents

535338

Sigma-Aldrich

2-Chloro-3-pyridinecarbonitrile

98%

Synonym(s):

2-Chloronicotinonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3ClN2
CAS Number:
Molecular Weight:
138.55
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

104-107 °C (lit.)

SMILES string

Clc1ncccc1C#N

InChI

1S/C6H3ClN2/c7-6-5(4-8)2-1-3-9-6/h1-3H

InChI key

JAUPUQRPBNDMDT-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katalin Nógrádi et al.
Bioorganic & medicinal chemistry letters, 24(16), 3845-3849 (2014-07-16)
An HTS campaign of our corporate compound library resulted in thieno[2,3-b]pyridines derivative hits with mGluR5 negative allosteric modulator effects. During the hit-to-lead development our objective was to improve affinity, and to keep the ligand efficiency values at an acceptable level.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service