Skip to Content
Merck
All Photos(1)

Key Documents

289833

Sigma-Aldrich

Strontium carbonate

≥98%

Synonym(s):

Strontianite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SrCO3
CAS Number:
Molecular Weight:
147.63
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Assay

≥98%

form

powder

density

3.7 g/mL at 25 °C (lit.)

SMILES string

[Sr++].[O-]C([O-])=O

InChI

1S/CH2O3.Sr/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

LEDMRZGFZIAGGB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Strontium carbonate is insoluble in water. It is used predominantly in producing other strontium salts.

Application

Strontium carbonate is used to prepare:
  • Barium strontium titanate
  • Barium strontium titanate thin films
It may be used to prepare europium doped di-strontium magnesium di-silicate phosphor.
Strontium carbonate is used in pyrotechnics, ceramic ferrites, inexpensive fireworks colorant.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photoluminescence properties of europium doped di-strontium magnesium di-silicate phosphor by solid state reaction method
Sahu IP, et al.
Science, 1-6 (2014)
Barium strontium titanate powder obtained by polymeric precursor method
Ries A, et al.
Materials Characterization, 50(2), 217-221 (2003)
Effect of process parameters and post deposition annealing on the optical, structural and microwave dielectric properties of RF magnetron sputtered (Ba 0.5, Sr 0.5) TiO 3 thin films
Saravanan K, et al.
Vacuum, 81(3), 307-316 (2006)
Debabrata Rautaray et al.
Langmuir : the ACS journal of surfaces and colloids, 20(16), 6827-6833 (2004-07-28)
The total biological synthesis of SrCO3 crystals of needlelike morphology arranged into higher order quasi-linear superstructures by challenging microorganisms such as fungi with aqueous Sr2+ ions is described. We term this procedure "total biological synthesis" since the source of carbonate
Wei Bo Li et al.
Radiation and environmental biophysics, 45(2), 115-124 (2006-05-31)
Intestinal absorption of strontium (Sr) in thirteen healthy adult German volunteers has been investigated by simultaneous oral and intravenous administration of two stable tracer isotopes, i.e. (84)Sr and (86)Sr. The measured Sr tracer concentration in plasma was analyzed using the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service