Skip to Content
Merck
All Photos(1)

Key Documents

277460

Sigma-Aldrich

3-(Methylthio)propionaldehyde

96%

Synonym(s):

3-(Methylmercapto)propionaldehyde, 3-(Methylthio)propanal, Methional, NSC 15874

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SCH2CH2CHO
CAS Number:
Molecular Weight:
104.17
Beilstein:
1739289
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

vapor pressure

760 mmHg ( 165 °C)

Assay

96%

form

liquid

refractive index

n20/D 1.483 (lit.)

bp

165-166 °C (lit.)

density

1.043 g/mL at 25 °C (lit.)

functional group

aldehyde

storage temp.

2-8°C

SMILES string

CSCCC=O

InChI

1S/C4H8OS/c1-6-4-2-3-5/h3H,2,4H2,1H3

InChI key

CLUWOWRTHNNBBU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-(Methylthio)propionaldehyde (methional) is the major product of chemical methionine conversion via Strecker degradation in food.

Application

3-(Methylthio)propionaldehyde has been used as a reactant in:
  • Oxidative coupling and chemoselective oxidation
  • Intermolecular alkyne hydroacylation reactions
  • Oxidation of sulfides to sulfoxides
It can also be used as a biosynthetic precursor of ethylene.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M M W Etschmann et al.
Applied microbiology and biotechnology, 80(4), 579-587 (2008-07-04)
Yeasts can convert amino acids to flavor alcohols following the Ehrlich pathway, a reaction sequence comprising transamination, decarboxylation, and reduction. The alcohols can be further derivatized to the acetate esters by alcohol acetyl transferase. Using L: -methionine as sole nitrogen
Ethylene formation from methional.
W A Pryor et al.
Biochemical and biophysical research communications, 81(2), 498-503 (1978-03-30)
A Escudero et al.
Journal of agricultural and food chemistry, 48(9), 4268-4272 (2000-09-20)
A young white wine that had undergone spontaneous oxidation and showed a strong off-flavor reminiscent of cooked vegetables was demixed by salting out to obtain an ethanolic extract that retained the off-flavor of the wine, as was demonstrated by sensory
F Amárita et al.
FEMS microbiology letters, 204(1), 189-195 (2001-10-30)
Lactic acid bacteria were screened for methional production from 4-methylthio-2-ketobutanoate. Only Lactococcus lactis IFPL730 produced high amounts of methional. It was demonstrated that production of this compound was an exclusively enzymatic reaction. The present work describes for the first time
M Murata et al.
Japanese journal of cancer research : Gann, 90(3), 268-275 (1999-06-08)
Adduct formation has been considered to be a major causal factor of DNA damage by carcinogenic heterocyclic amines. By means of experiments with 32P-labeled DNA fragments and an electrochemical detector coupled to a high-pressure liquid chromatograph, we investigated whether the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service