Skip to Content
Merck
All Photos(2)

Key Documents

667420

Sigma-Aldrich

1H,1H,2H,2H-Perfluorooctyltriethoxysilane

98%

Synonym(s):

Triethoxy(1H,1H,2H,2H-perfluoro-1-octyl)silane, Triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-1-octyl)silane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19F13O3Si
CAS Number:
Molecular Weight:
510.36
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

refractive index

n20/D 1.344

density

1.3299 g/mL at 25 °C

SMILES string

CCO[Si](CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)(OCC)OCC

InChI

1S/C14H19F13O3Si/c1-4-28-31(29-5-2,30-6-3)8-7-9(15,16)10(17,18)11(19,20)12(21,22)13(23,24)14(25,26)27/h4-8H2,1-3H3

InChI key

AVYKQOAMZCAHRG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1H,1H,2H,2H-Perfluorooctyltriethoxysilane(FOTS) is a fluorinated alkyl silane that is majorly used for improving the wettability of the substrate by lowering the surface energy. It facilitates a superhydrophobic coating by enriching the surface with CF3 group and a water contact angle above 150°.
Triethoxy(1H,1H,2H,2H-perfluoro-1-octyl)silane (POTS) is a fluorine based polymer. POTS possesses low surface free energy leading to its anti-adhesive behavior to polar and non-polar substances. It shows very good weathering stability on account of the carbon- fluorine bond. POTS hydrolyzes in wet environment to form silane based film and self healing anticorrosive polymers, the film formed from the hydrolysis and polycondensation of POTS and water is hydrophobic in nature. This wetting property acts in repelling aqueous electrolyte solution away from metallic substrates and hence providing corrosion protection to metal substrate.

Application

POTS can be used as a silane based coating on poly(vinylidene fluoride) (PVDF) to synthesize a membrane for the removal of hazardous volatile organic compounds. Glass substrates can be modified with POTS to increase the contact angle for 3D reactive inkjet printing of polydimethylsiloxane (PDMS). It can also be used in the synthesis of superhydrophobic carbon nanotubes(CNT) hollow membranes for membrane desalination (MD) as eco-friendly products of fresh water. It is deposited on the roughened surface of titanium to render it superhydrophobic. POTS may be used as a component of fluorine modified ORMOCER (Organically Modified Ceramics) coating materials.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhancing membrane performance in removal of hazardous VOCs from water by modified fluorinated PVDF porous material.
Al-Gharabli S, et al.
Journal of Membrane Science , 556, 214-226 (2018)
Lin Wang et al.
Science advances, 6(29), eabb2307-eabb2307 (2020-08-25)
Many natural surfaces are capable of rapidly shedding water droplets-a phenomenon that has been attributed to the presence of low solid fraction textures (Φs ~ 0.01). However, recent observations revealed the presence of unusually high solid fraction nanoscale textures (Φs
Fabricating superhydrophobic and oleophobic surface with silica nanoparticles modified by silanes and environment-friendly fluorinated chemicals.
Cai Y, et al.
Applied Surface Science, 450, 102-111 (2018)
Zhongshun Wang et al.
Nanoscale, 9(43), 16749-16754 (2017-10-27)
The performance of surface-enhanced Raman scattering (SERS) for detecting trace amounts of analytes depends highly on the enrichment of the diluted analytes into a small region that can be detected. A super-hydrophobic delivery (SHD) process is an excellent process to
Xiang Li et al.
ACS applied materials & interfaces, 10(14), 12042-12050 (2018-03-21)
Electrically conductive fabrics with liquid repellency and corrosive resistance are strongly desirable for wearable displays, biomedical sensors, and so forth. In the present work, highly electrically conductive and healable superamphiphobic cotton fabrics are fabricated by a solution-dipping method that involves

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service