Skip to Content
Merck
All Photos(1)

Key Documents

251178

Sigma-Aldrich

1,6-Diiodohexane

97%, contains copper as stabilizer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
I(CH2)6I
CAS Number:
Molecular Weight:
337.97
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.5852 (lit.)

bp

141-142 °C/10 mmHg (lit.)

mp

9-10 °C (lit.)

density

2.05 g/mL at 25 °C (lit.)

functional group

iodo

SMILES string

ICCCCCCI

InChI

1S/C6H12I2/c7-5-3-1-2-4-6-8/h1-6H2

InChI key

QLIMAARBRDAYGQ-UHFFFAOYSA-N

Application

1,6-Diiodohexane has been used:
  • in preparation of cationic tissue plasminogen activator (t-PA)-containing polyurethane surfaces
  • as solvent additive, to improve the power conversion effciency of polymer/fullerene bulk heterojunction solar cells
  • synthesis of new macrocycles, having 2,4-dihydroxybenzophenone as a connecting unit
  • preparation of a novel ionic electrolyte, 3-(iodohexyl)-1-(3-(triethoxysilyl)propylcarbamoyl)-1H-benzo[d]imidazol-3-ium iodide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and structure of new cyclophanes containing benzofuran and benzene rings.
Park KK, et al.
Tetrahedron Letters, 41(9), 1393-1396 (2000)
Novel silane-substituted benzimidazolium iodide as gel electrolyte for dye-sensitized solar cells.
Lee JP, et al.
Electrochimica Acta, 54(18), 4635-4670 (2009)
Zhaoqiang Wu et al.
Acta biomaterialia, 7(5), 1993-1998 (2011-01-25)
With the aim of minimizing thrombus formation in blood-contacting devices, tissue plasminogen activator (t-PA)-containing polyurethane (PU) materials have been developed. Cationic PU surfaces were prepared by grafting poly(dimethylaminoethyl methacrylate) and quaternizing the tertiary amino groups with iodomethane or 1,6-diiodohexane or
Improving device efficiency of polymer/fullerene bulk heterojunction solar cells through enhanced crystallinity and reduced grain boundaries induced by solvent additives.
Ming-Shin Su et al.
Advanced materials (Deerfield Beach, Fla.), 23(29), 3315-3319 (2011-06-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service