Skip to Content
Merck
All Photos(3)

Key Documents

236330

Sigma-Aldrich

p-Toluenesulfonamide

ReagentPlus®, ≥99%

Synonym(s):

4-Methylbenzene-1-sulfonamide, p-Tosylamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2NH2
CAS Number:
Molecular Weight:
171.22
Beilstein:
472689
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
≥99%
Pricing and availability is not currently available.

Quality Level

product line

ReagentPlus®

Assay

≥99%

form

solid

mp

134-137 °C (lit.)

SMILES string

Cc1ccc(cc1)S(N)(=O)=O

InChI

1S/C7H9NO2S/c1-6-2-4-7(5-3-6)11(8,9)10/h2-5H,1H3,(H2,8,9,10)

InChI key

LMYRWZFENFIFIT-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Cyanomethylenetributylphosphorane (CMBP)-mediated reaction of p-toluenesulfonamide with alcohols has been investigated[1].

Application

p-Toluenesulfonamide has been employed:
  • as nucleophile during tetrabutylammonium fluoride (TBAF) catalyzed vinyl aziridine opening reaction[2]
  • as reagent during selective aziridination of olefins catalyzed by dirhodium (II) caprolactamate[3]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

395.6 °F - closed cup

Flash Point(C)

202 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Simone et al.
Lab on a chip, 18(15), 2235-2245 (2018-06-28)
In this work, a novel multi-microfluidic crystallization platform called MMicroCryGen is presented, offering a facile methodology for generating individual crystals for fast and easy screening of the polymorphism and crystal habit of solid compounds. The MMicroCryGen device is capable of
Mitsunobu-type alkylation of p-toluenesulfonamide. A convenient new route to primary and secondary amines.
Tsunoda T, et al.
Tetrahedron Letters, 37(!4), 2457-2458 (1996)
Arthur J Catino et al.
Organic letters, 7(13), 2787-2790 (2005-06-17)
[reaction: see text] A mild, efficient, and selective aziridination of olefins catalyzed by dirhodium(II) caprolactamate [Rh(2)(cap)(4).2CH(3)CN] is described. Use of p-toluenesulfonamide (TsNH(2)), N-bromosuccinimide (NBS), and potassium carbonate readily affords aziridines in isolated yields of up to 95% under extremely mild
Charles H Reynolds et al.
Journal of medicinal chemistry, 51(8), 2432-2438 (2008-04-03)
Ligand efficiency (i.e., potency/size) has emerged as an important metric in drug discovery. In general, smaller, more efficient ligands are believed to have improved prospects for good drug properties (e.g., bioavailability). Our analysis of thousands of ligands across a variety
An efficient FeCl3 -catalyzed amidation reaction of secondary benzylic and allylic alcohols with carboxamides or p-toluenesulfonamide.
Jana U, et al.
Tetrahedron Letters, 42(37), 6433-6435 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service