Skip to Content
Merck
All Photos(2)

Key Documents

246867

Sigma-Aldrich

2-Cyclopentylphenol

technical grade, 95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H9C6H4OH
CAS Number:
Molecular Weight:
162.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

95%

form

solid

refractive index

n20/D 1.555 (lit.)

bp

148-150 °C/18 mmHg (lit.)

mp

34-35 °C (lit.)

SMILES string

Oc1ccccc1C2CCCC2

InChI

1S/C11H14O/c12-11-8-4-3-7-10(11)9-5-1-2-6-9/h3-4,7-9,12H,1-2,5-6H2

InChI key

JHEKSKQMOBLXQS-UHFFFAOYSA-N

Gene Information

human ... GABRA1(2554)

Application

2-Cyclopentylphenol has been used in the synthesis of:
  • optically active (S)-penbutolol (antihypertensive drug) via Sharpless asymmetric dihydroxylation
  • (S)-1-(2,3-epoxypropoxy)-2-chloro-5-methyl-benzene

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regioselective alkylation of phenol with cyclopentanol over montmorillonite K10: An efficient synthesis of 1-(2-cyclopentylphenoxy)-3-[(1, 1-dimethylethyl) amino] propan-2-ol {(S)-penbutolol}.
Phukan P and Sudalai A.
Journal of the Chemical Society. Perkin Transactions 1, 20, 3015-3018 (1999)
Petra Doze et al.
Neurochemistry international, 40(2), 145-155 (2001-12-12)
Five potent, lipophilic beta-adrenoceptor antagonists (carvedilol, pindolol, toliprolol and fluorinated analogs of bupranolol and penbutolol) were labeled with either carbon-11 or fluorine-18 and evaluated for cerebral beta-adrenoceptor imaging in experimental animals. The standard radioligand for autoradiography of beta-adrenoceptors, [125I]-iodocyanopindolol, was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service