Skip to Content
Merck
All Photos(2)

Documents

236616

Sigma-Aldrich

1,2,4,5-Tetrakis(bromomethyl)benzene

95%

Synonym(s):

α,α′,α′′,α′′′-Tetrabromodurene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H2(CH2Br)4
CAS Number:
Molecular Weight:
449.80
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

159-161 °C (lit.)

SMILES string

BrCc1cc(CBr)c(CBr)cc1CBr

InChI

1S/C10H10Br4/c11-3-7-1-8(4-12)10(6-14)2-9(7)5-13/h1-2H,3-6H2

InChI key

UTXIKCCNBUIWPT-UHFFFAOYSA-N

Application

1,2,4,5-Tetrakis(bromomethyl)benzene has been used in the synthesis of:
  • tetrapodal imidazolium ligands as their PF6 salts
  • 6,7-bis(bromomethyl)-2,11,18,21,24-pentaoxatetracyclo[23.4. 0.04,9.012,17]nonacosa-1(25),4(9),5,7,12 (17),13,15,26,28-nonaene
  • conformationally constrained cyclic α-amino acid derivatives
  • thiacyclophanes
  • dendrimeric organoplatinum complexes

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Canadian Journal of Chemistry, 72, 1728-1728 (1994)
B Nisar Ahamed et al.
Inorganic chemistry, 52(8), 4269-4276 (2013-03-26)
Tetrapodal imidazolium ligands L(1)-L(3) as their PF6(-) salts are synthesized in good yields by reacting 1,2,4,5-tetrakis(bromomethyl)benzene with N-methylimidazole, N-benzylimidazole, and N-ethylimidazole, respectively. Single-crystal X-ray diffraction studies of L(1)·4PF6, L(2)·4PF6, and L(3)·4PF6 show the chair conformation of the tetrapodal imidazoliums (L(1)-L(3))
W Sim et al.
Acta crystallographica. Section C, Crystal structure communications, 57(Pt 3), 293-294 (2001-03-16)
The 17-crown-5 unit, C(26)H(26)Br(2)O(5), consisting of a 1,2-bis(bromomethyl) group, three benzo groups and diethylene glycol, was prepared from the reaction of 1,2,4,5-tetrakis(bromomethyl)benzene and bis-phenol in the presence of sodium hydride as a base. This molecule seems to offer an internal
Journal of the Chemical Society. Chemical Communications, 1895-1895 (1994)
Journal of the Chemical Society. Perkin Transactions 1, 1883-1883 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service