Skip to Content
Merck
All Photos(2)

Key Documents

D204803

Sigma-Aldrich

Diphenylacetylene

98%

Synonym(s):

Tolan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C≡CC6H5
CAS Number:
Molecular Weight:
178.23
Beilstein:
606478
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

crystals

bp

170 °C/19 mmHg (lit.)

mp

59-61 °C (lit.)

density

0.99 g/mL at 25 °C (lit.)

SMILES string

c1ccc(cc1)C#Cc2ccccc2

InChI

1S/C14H10/c1-3-7-13(8-4-1)11-12-14-9-5-2-6-10-14/h1-10H

InChI key

JRXXLCKWQFKACW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noni K Gaylord-Harden et al.
The American journal of orthopsychiatry, 87(4), 463-473 (2016-12-16)
The current study examined pathways in a model of desensitization, the Pathologic Adaptation Model, in adolescent males of color. Specifically, the current study examined depressive symptoms and deviant beliefs as mediators of the association between community violence exposure and subsequent
Hongmei Liu et al.
The Journal of chemical physics, 129(22), 224704-224704 (2008-12-17)
A series of linear conjugated molecular wires (diphenylacetylene connected by double or triple bonds) asymmetrically substituted by various functional groups was investigated by using density functional theory combined with nonequilibrium Green's function method. The transportation behaviors of these models did
Wenjing Hong et al.
Journal of the American Chemical Society, 134(4), 2292-2304 (2011-12-20)
Employing a scanning tunneling microscopy based beak junction technique and mechanically controlled break junction experiments, we investigated tolane (diphenylacetylene)-type single molecular junctions having four different anchoring groups (SH, pyridyl (PY), NH(2), and CN) at a solid/liquid interface. The combination of
Rajesh Chandra et al.
Journal of medicinal chemistry, 50(10), 2415-2423 (2007-04-24)
A series of 18F fluoropegylated diphenylacetylenes as probes for binding to Abeta plaques were successfully prepared. These relatively rigid acetylenes, 12a, 12b, 14a, and 14b, displayed high binding affinities in postmortem AD brain homogenates (Ki ranging from 1.2 to 2.9
Jim C Spain et al.
Applied and environmental microbiology, 69(7), 4037-4042 (2003-07-04)
Several strategies for using enzymes to catalyze reactions leading to the synthesis of relatively simple substituted picolinic acids have been described. The goal of the work described here was to synthesize a more complex molecule, 6-phenylacetylene picolinic acid [6-(2-phenylethynyl)pyridine-2-carboxylic acid]

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service