Skip to Content
Merck
All Photos(2)

Documents

219797

Sigma-Aldrich

2-Hydroxybenzotrifluoride

97%

Synonym(s):

α,α,α-Trifluoro-o-cresol, 2-Trifluoromethylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4OH
CAS Number:
Molecular Weight:
162.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

bp

147-148 °C (lit.)

mp

45-46 °C (lit.)

SMILES string

Oc1ccccc1C(F)(F)F

InChI

1S/C7H5F3O/c8-7(9,10)5-3-1-2-4-6(5)11/h1-4,11H

InChI key

ZOQOPXVJANRGJZ-UHFFFAOYSA-N

Application

2-Hydroxybenzotrifluoride was used to investigate the self-aggregation of polyanion-containing cinnamoyl group with diazoresin (DR) to form stable covalent ultrathin film by UV irradiation.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photoalignment of liquid crystals by a covalently attached self-assembled ultrathin film.
Zhang L, et al.
Crystals, 39(8), 983-991 (2012)
Douglas H Read et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 35(6), 671-677 (2019-02-19)
We describe for the first time hydrogen bonded acid (HBA) polymer, poly{methyl[3-(2-hydroxyl, 4,6-bistrifluoromethyl)phenyl]propylsiloxane}, (DKAP), as stationary phase for gas chromatography (μGC) of organophosphate (OP), chemical warfare agent (CWA) surrogates, dimethylmethylphosphonate (DMMP), diisopropylmethylphosphonate (DIMP), diethylmethylphosphonate (DEMP), and trimethylphosphate (TMP), with high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service