201677
Janus Green B
certified by the Biological Stain Commission, Dye content 65 %
Synonym(s):
3-Diethylamino-7-(4-dimethylaminophenylazo)-5-phenylphenazinium chloride, Diazin Green S, Union Green B
About This Item
Recommended Products
grade
certified by the Biological Stain Commission
Quality Level
form
powder
composition
Dye content, 65%
color
black
very dark brown
very dark green
λmax
395 nm
660 nm (2nd)
ε (extinction coefficient)
≥28000 at 651-677 nm in ethanol: water (1:1) at 0.01 g/L
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
2-8°C
SMILES string
[Cl-].CCN(CC)c1ccc2nc3ccc(cc3[n+](-c4ccccc4)c2c1)\N=N\c5ccc(cc5)N(C)C
InChI
1S/C30H31N6.ClH/c1-5-35(6-2)26-17-19-28-30(21-26)36(25-10-8-7-9-11-25)29-20-23(14-18-27(29)31-28)33-32-22-12-15-24(16-13-22)34(3)4;/h7-21H,5-6H2,1-4H3;1H/q+1;/p-1
InChI key
XXACTDWGHQXLGW-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Application
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service