192023
Poly(acrylic acid) partial sodium salt solution
average Mw ~2,000 by GPC, 61-65 wt. % in H2O
Synonym(s):
Acrylic acid-sodium acrylate copolymer, Sodium acrylate-acrylic acid copolymer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
mol wt
average Mw ~2,000 by GPC
composition
Na, 0.6%
concentration
61-65 wt. % in H2O
refractive index
n20/D 1.442
density
1.2 g/mL at 25 °C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Sodium salt of poly(acrylic acid) is a superabsorbent polymer. Superabsorbents are composed of crosslinked networks of hydrophilic polymers. It can absorb and retain a large capacity of water based fluids, the release of the absorbed fluids is tough even under an applied pressure. Absorption capacity is inversely proportional to salt concentration and applied load.They are formed via solution or inverse-suspension polymerization techniques. †
Application
Sodium salt of poly (acrylic acid) may be used as a major component of non glare coatings for headlights.It acts as an antiscaling agents.It′s effect of the salt on the crystal (Cu and MS metals) formation was studied. General applications of these superabsorbents are in many fields such as agriculture and horticulture, sanitary goods, sealing composites, component of moisture barrier tapes and coatings, humidity control, gel actuators. and medicine for drug-delivery systems.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Synthesis and characterization of chitosan-g-poly (acrylic acid)/attapulgite superabsorbent composites
Zhang J, et al.
Carbohydrate Polymers, 68(2), 367-374 null
Study on Na and K salts of PAA as corrosion inhibitors
Mishra S, et al.
J. Sci. Ind. Res., 64, 684-687 (2005)
Superabsorbent Polymer Materials: A Review
Zohuriaan-Mehr MJ and Kabiri K
Iranian Polymer Journal, 17(6), 451-477 (2008)
Studies on poly (acrylic acid)/attapulgite superabsorbent composite. I. Synthesis and characterization
Li An, et al.
Journal of Applied Polymer Science, 92(3), 1596-1603 (2004)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service