Skip to Content
Merck
All Photos(3)

Key Documents

447013

Sigma-Aldrich

Poly(acrylic acid sodium salt)

average Mw ~5,100 by GPC, powder

Synonym(s):

Poly(sodium acrylate), Sodium polyacrylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

form

powder

Quality Level

mol wt

average Mw ~5,100 by GPC

impurities

8-14% water

pH

6-9

density

0.55 g/mL at 25 °C

SMILES string

[Na+].[O-]C(=O)C=C

InChI

1S/C3H4O2.Na/c1-2-3(4)5;/h2H,1H2,(H,4,5);/q;+1/p-1

InChI key

NNMHYFLPFNGQFZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Scale inhibitor, builder in detergents, aid in soap and detergent processing and control of water hardness.

Features and Benefits

Water soluble sequesters metal ions and hydrolytically and thermally stable.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service