Skip to Content
Merck
All Photos(1)

Key Documents

186481

Sigma-Aldrich

6-Bromo-1-hexanol

97%

Synonym(s):

Hexamethylene bromohydrin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)6OH
CAS Number:
Molecular Weight:
181.07
Beilstein:
1732415
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.482 (lit.)

bp

105-106 °C/5 mmHg (lit.)

density

1.384 g/mL at 25 °C (lit.)

functional group

bromo
hydroxyl

storage temp.

2-8°C

SMILES string

BrCCCCCCO

InChI

1S/C6H13BrO/c7-5-3-1-2-4-6-8/h8H,1-6H2

InChI key

FCMCSZXRVWDVAW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

6-Bromo-1-hexanol, also known as hexamethylene bromohydrin is a cross-linking reagent that is commonly used in organic and polymeric synthesis.

Application

6-Bromo-1-hexanol is used as a cross linker to synthesize cyclic, branched, and bicyclic oligonucleotides by click chemistry. It is also utilized to synthesize gel polymer electrolytes, and polymerizable ionic liquids (PILs) such as 1-[(6-methacryloyloxy)hexyl]3-methylimidazolium bromide.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Polymerizable Ionic Liquid Monomer and Its Characterization
J Sardar
IOP Conference Series: Materials Science and Engineering, 111, 012021-012021 (2016)
Cycling performance of lithium polymer cells assembled by in situ polymerization of a non-flammable ionic liquid monomer
YS Lee
Electrochimica Acta, 106, 460-464 (2013)
New strategies for cyclization and bicyclization of oligonucleotides by click chemistry assisted by microwaves
J Lietard
The Journal of Organic Chemistry, 73, 191-200 (2008)
Zrinka Rajic et al.
Journal of inorganic biochemistry, 169, 50-60 (2017-01-29)
We disclose here the studies that preceded and guided the preparation of the metal-based, redox-active therapeutic Mn(III) meso-tetrakis(N-n-butoxyethylpyridyl)porphyrin, MnTnBuOE-2-PyP
Eike T Röchow et al.
Polymers, 12(8) (2020-08-06)
Solid polymer electrolytes for bipolar lithium ion batteries requiring electrochemical stability of 4.5 V vs. Li/Li+ are presented. Thus, imidazolium-containing poly(ionic liquid) (PIL) networks were prepared by crosslinking UV-photopolymerization in an in situ approach (i.e., to allow preparation directly on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service