Skip to Content
Merck
All Photos(1)

Key Documents

160415

Sigma-Aldrich

3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine

≥99%

Synonym(s):

5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine, PDT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14N4
CAS Number:
Molecular Weight:
310.35
Beilstein:
542151
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

mp

191-193 °C (lit.)

functional group

phenyl

SMILES string

c1ccc(cc1)-c2nnc(nc2-c3ccccc3)-c4ccccn4

InChI

1S/C20H14N4/c1-3-9-15(10-4-1)18-19(16-11-5-2-6-12-16)23-24-20(22-18)17-13-7-8-14-21-17/h1-14H

InChI key

OTMYLOBWDNFTLO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine (PDT) reacts with Cu(II) perchlorate in ethanol to yield [Cu(PDT)2(ClO4)2].

Application

3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine was employed as chromogenic-extraction reagent during spectrophotometric determination of iron in acids and acidic solutions. It was also employed as precolumn derivatizing reagent in the HPLC method with UV absorbance detection for the Fe(II) determination.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Spectrophotometric determination of iron in acids and acidic solutions by an extraction formation reaction involving 3-(2-pyridyl)-5, 6-diphenyl-1, 2, 4-triazine as the chromogenic-extraction reagent.
Chriswell CD and Schilt AA.
Analytical Chemistry, 46(8), 992-996 (1974)
Synthesis and structural charaterization of a Cu (II) complex with 3-(2-pyridyl)-5, 6-diphenyl-1, 2, 4-triazine (PDPT).
Soudi AA, et al.
Journal of Coordination Chemistry, 58(17), 1631-1637 (2005)
Yu-Cherng Channing Chang et al.
Scientific reports, 7(1), 9746-9746 (2017-08-31)
Tumor heterogeneity can be elucidated by mapping subregions of the lesion with differential imaging characteristics, called habitats. Dynamic Contrast Enhanced (DCE-)MRI can depict the tumor microenvironments by identifying areas with variable perfusion and vascular permeability, since individual tumor habitats vary
Thomas J van Rijssen et al.
PloS one, 13(8), e0202549-e0202549 (2018-08-25)
To assess parameters on optical coherence tomography (OCT), and their correlation with best-corrected visual acuity (BCVA) in patients with non-resolving central serous chorioretinopathy (CSC). For 25 non-resolving CSC patients treated with photodynamic therapy (PDT), the thickness of retinal layers was
Yvonne H W Derks et al.
Theranostics, 11(4), 1527-1541 (2021-01-08)
Incomplete resection of prostate cancer (PCa) occurs in 15%-50% of PCa patients. Disease recurrence negatively impacts oncological outcome. The use of radio-, fluorescent-, or photosensitizer-labeled ligands to target the prostate-specific membrane antigen (PSMA) has become a well-established method for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service