Recommended Products
Assay
99%
form
solid
mp
158-160 °C (lit.)
SMILES string
[O-][N+](=O)c1ccc(\C=C\C(=O)c2ccccc2)cc1
InChI
1S/C15H11NO3/c17-15(13-4-2-1-3-5-13)11-8-12-6-9-14(10-7-12)16(18)19/h1-11H/b11-8+
InChI key
WDZGGAFMGIOIQS-DHZHZOJOSA-N
General description
Mutagenic activity of 4-nitrochalcone on two strains of Salmonella typhimurium (TA98 and TA100) with and without rat liver microsomal and cytosolic enzymes has been tested.
Application
4-Nitrochalcone was used in the synthesis of 2,3-dibromo-4-nitrochalcone.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis and Photochromic Properties of New Heterocyclic Derivatives of 1, 3-Diazabicyclo [3.1. 0] Hex-3-Ene.
J. Chin. Chem. Soc., 54(3), 635-641 (2007)
European journal of pharmacology, 884, 173392-173392 (2020-08-01)
The Leishmaniasis treatment currently available involves some difficulties, such as high toxicity, variable efficacy, high cost, therefore, it is crucial to search for new therapeutic alternatives. Over the past few years, research on new drugs has focused on the use
Mutation research, 169(3), 71-79 (1986-03-01)
31 p-monosubstituted chalcones (E-1, 3-diphenylpropene-1-one) and the corresponding oxides (E-1-benzoyl-2-phenyloxirane) were tested for mutagenic activity on two strains of Salmonella typhimurium (TA98 and TA100) with and without rat liver microsomal and cytosolic enzymes. Highest mutagenicity (3.0 revertants/nmole in either strain)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service