Skip to Content
Merck
All Photos(1)

Documents

558311

Sigma-Aldrich

4-Chlorochalcone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH=CHOCOC6H5
CAS Number:
Molecular Weight:
242.70
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

113-117 °C (lit.)

SMILES string

Clc1ccc(cc1)\C=C\C(=O)c2ccccc2

InChI

1S/C15H11ClO/c16-14-9-6-12(7-10-14)8-11-15(17)13-4-2-1-3-5-13/h1-11H/b11-8+

InChI key

ABGIIXRNMHUKII-DHZHZOJOSA-N

General description

4-Chlorochalcone is a chalcone derivative. Its crystals exhibit monoclinic system with space group P21/c. It can undergo conjugate addition reaction with pyrrole in the presence of AlKIT-5 (mesoporous 3D aluminosilicate catalyst) to form the corresponding C2-alkylated pyrrole derivatives.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly Efficient Friedel-Crafts Alkylation of Indoles and Pyrrole Catalyzed by Mesoporous 3D Aluminosilicate Catalyst with Electron-Deficient Olefins.
Naidu TS, et al.
Synlett, 18, 2813-2817 (2010)
Structure of 4-chlorochalcone.
Li Z and Su G.
Acta Crystallographica Section C, Crystal Structure Communications, 50(1), 126-127 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service