Skip to Content
Merck
All Photos(1)

Key Documents

149101

Sigma-Aldrich

9-Aminophenanthrene

96%

Synonym(s):

9-Phenanthrenamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11N
CAS Number:
Molecular Weight:
193.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

mp

137-139 °C (lit.)

SMILES string

Nc1cc2ccccc2c3ccccc13

InChI

1S/C14H11N/c15-14-9-10-5-1-2-6-11(10)12-7-3-4-8-13(12)14/h1-9H,15H2

InChI key

KIHQWOBUUIPWAN-UHFFFAOYSA-N

Application

9-Aminophenanthrene was used as a model ligand to study the binding of 9-aminophenanthrene to individual subsites within the active site of CYP(eryF).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Y Aboul-Enein et al.
The Journal of pharmacy and pharmacology, 50(3), 291-296 (1998-05-26)
Racemic ketoprofen is a non-steroidal anti-inflammatory drug used to treat musculoskeletal and colic conditions in horses. The enantioselective chiral inversion of ketoprofen administered to horses has been studied by use of cellulose tris(4-methylbenzoate), also known as Chiralcel OJ-R, as chiral
Arthur G Roberts et al.
Biochemistry, 45(6), 1673-1684 (2006-02-08)
Cytochrome P450's (P450's) catalyze the oxidative metabolism of most drugs and toxins. Although extensive studies have proven that some P450's demonstrate both homotropic and heterotropic cooperativity toward a number of substrates, the mechanistic and molecular details of P450 allostery are
Kishore K Khan et al.
Chemical research in toxicology, 15(6), 806-814 (2002-06-18)
The main objective of the present study was to find a fluorescent substrate probe for cytochrome P450eryF (P450eryF). P450eryF is a bacterial P450 that catalyzes the hydroxylation of 6-deoxyerythronolide B at the 6S position, a necessary step in the biosynthesis
M Ikeda et al.
Journal of chromatography, 305(2), 261-270 (1984-02-10)
The application of 9-aminophenanthrene (9-AP), a fluorescence-labeling reagent for free fatty acids (FFA), was examined. 9-AP dissolved in benzene was added to a benzene solution of FFA chlorides derived from FFA and oxalyl chloride. The mixture was allowed to react
Moon-Young Yoon et al.
Drug metabolism reviews, 36(2), 219-230 (2004-07-09)
Cytochrome P450-dependent drug metabolism in vitro frequently deviates from simple Michaelis-Menten kinetic models, and demonstrates both positive and negative homotropic and heterotropic effects. These complex "allsoteric" kinetics confound our ability to predict drug clearance, and they may provide a basis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service