Skip to Content
Merck
All Photos(1)

Key Documents

139440

Sigma-Aldrich

Diethyl 2-bromo-2-methylmalonate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC(CH3)(CO2C2H5)2
CAS Number:
Molecular Weight:
253.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.449 (lit.)

density

1.325 g/mL at 25 °C (lit.)

functional group

bromo
ester

SMILES string

CCOC(=O)C(C)(Br)C(=O)OCC

InChI

1S/C8H13BrO4/c1-4-12-6(10)8(3,9)7(11)13-5-2/h4-5H2,1-3H3

InChI key

CSLQAXTUGPUBCW-UHFFFAOYSA-N

General description

Diethyl 2-bromo-2-methylmalonate acts as initiator during the atom transfer radical polymerization of methyl methacrylate.

Application

Diethyl 2-bromo-2-methylmalonate was used to investigate the 1,2,2,6,6-pentamethylpiperidine as an effective reducing agent for the radical chain conversion of primary bromoesters to the corresponding esters. It was used in the synthesis of polystyrene macroinitiator via atom transfer radical polymerization.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TAILOR MADE POLYMERIC SYSTEMATIC LIBRARIES VIA ATOM TRANSFER RADICAL POLYMERIZATION.
Jakubowski W, et al.
Polymer Preprints (American Chemical Society, Division of Polymer Chemistry), 49(2), 369-369 (2008)
Organic reducing agents. Reduction of electron deficient bromides by 1, 2, 2, 6, 6-pentamethylpiperidine (PMP)/mercaptoethanol.
Amoli M, et al.
Tetrahedron Letters, 36(23), 3997-4000 (1995)
Controlled/?living? atom transfer radical polymerization of methyl methacrylate using various initiation systems.
Matyjaszewski K, et al.
Macromolecules, 31(5), 1527-1534 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service