Skip to Content
Merck
All Photos(1)

Documents

132756

Sigma-Aldrich

1,2,4-Tribromobenzene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3Br3
CAS Number:
Molecular Weight:
314.80
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

41-43 °C (lit.)

SMILES string

Brc1ccc(Br)c(Br)c1

InChI

1S/C6H3Br3/c7-4-1-2-5(8)6(9)3-4/h1-3H

InChI key

FWAJPSIPOULHHH-UHFFFAOYSA-N

General description

1,2,4-tribromobenzene on photochemical dehalogenation in open-air solutions of acetonitrile yields 1,4-dibromobenzene, 1,3-dibromobenzene and 1,2-dibromobenzene.

Application

1,2,4-tribromobenzene was used in the synthesis of hyperbranched poly(p-phenylene ethynylenes). It was used as cross-linking reagent during Pd-catalyzed cross-coupling of 2,5-diiodo-4-[(2-ethylhexyl)oxy]methoxybenzene and 1,4-diethynyl-2,5-bis-(octyloxy)benzene.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyperbranched Poly (p-phenylene ethynylene) s.
Mendez JD, et al.
Macromolecular Chemistry and Physics, 208(15), 1625-1636 (2007)
E N Smith et al.
Journal of toxicology and environmental health, 6(4), 737-749 (1980-07-01)
1,2,4-Trichlorobenzene (TCB) and 1,2,4-tribromobenzene (TBB) were administered for 7 d to rats at a dose of 1 mmol/kg.d. The animals were sacrificed at various times to observe the decline in enzyme induction. Carbon 14-labeled TCB and TBB were administered and
Synthesis and characterization of cross-linked conjugated polymer milli-, micro-, and nanoparticles.
Eric Hittinger et al.
Angewandte Chemie (International ed. in English), 43(14), 1808-1811 (2004-04-01)
P K Freeman et al.
Journal of chemical technology and biotechnology (Oxford, Oxfordshire : 1986), 72(1), 45-49 (1998-06-17)
The photochemical dehalogenation of 1,2,4-tribromobenzene, 1,2,3,5-tetrabromobenzene and pentachlorobenzene in open-air solutions of acetonitrile using natural and artificial sunlight as the irradiation source has been investigated. The regiochemistry of mono-dehalogenation has been determined for 1,2,4-tribromobenzene and 1,2,3,5-tetrabromobenzene. Pentachlorobenzene did not react.
Darol E Dodd et al.
International journal of toxicology, 31(3), 250-256 (2012-04-06)
Male Sprague-Dawley rats were exposed to 1,2,4-tribromobenzene (TBB) by gavage for 5 days, 2, 4, and 13 weeks at 0, 2.5, 5, 10, 25, or 75 mg/kg per d. There were no TBB exposure-related clinical signs of toxicity or changes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service