Skip to Content
Merck
All Photos(1)

Key Documents

107131

Sigma-Aldrich

Hexabromobenzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6Br6
CAS Number:
Molecular Weight:
551.49
Beilstein:
1912586
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

>300 °C (lit.)

solubility

chloroform: soluble 10 mg/mL

functional group

bromo

SMILES string

Brc1c(Br)c(Br)c(Br)c(Br)c1Br

InChI

1S/C6Br6/c7-1-2(8)4(10)6(12)5(11)3(1)9

InChI key

CAYGQBVSOZLICD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexabromobenzene is used as fire retardant in plastics, paper and electric manufactured goods. It undergoes protodebromination when treated with sodium methoxide in methanol and ethyl methyl ketone to give a mixture of tetrabromobenzenes.

Application

The porphyrinogenic potential of hexabromobenzene (HBB) with hexachlorobenzene was studied in the liver of primiparous rats.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Uptake of chloro- and bromobiphenyls, hexachloro- and hexabromobenzene by fish.
V Zitko et al.
Bulletin of environmental contamination and toxicology, 16(6), 665-673 (1976-12-01)
Polyhalogeno-aromatic compounds. Part VIII. Reactions of hexabromobenzene with nucleophiles.
Collins I and Suschitzky H.
J. Chem. Soc. Sect. C, 18, 2337-2341 (1969)
Comparison of the porphyrinogenic activity of hexabromobenzene and hexachlorobenzene in primiparous Wistar rats.
C E Mendoza et al.
Bulletin of environmental contamination and toxicology, 21(3), 358-364 (1979-02-01)
Hans Peter H Arp et al.
Journal of environmental monitoring : JEM, 13(3), 505-513 (2010-12-09)
The brominated flame retardants (BFRs), pentabromotoluene (PBT), pentabromoethylbenzene (PBEB) and hexabromobenzene (HBB), exhibit physical-chemical properties similar to other persistent organic pollutants, and have been in use as flame retardants for several decades. Data on these BFRs in diverse environmental samples
A G Smith et al.
Research communications in chemical pathology and pharmacology, 28(2), 377-384 (1980-05-01)
Virgin female Agus rats were fed a diet containing hexahalogenated benzenes (700 nmol/g of food) for 70 days. The liver concentrations of these compounds were then determined and correlated with any depression in uroporphyrinogen decarboxylase activity and increases in porphyrin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service