Skip to Content
Merck
All Photos(4)

Documents

125237

Sigma-Aldrich

4-Chlorothiophenol

97%

Synonym(s):

4-Chlorobenzenethiol, 4-Chlorophenyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4SH
CAS Number:
Molecular Weight:
144.62
Beilstein:
605971
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

bp

205-207 °C (lit.)

mp

49-51 °C (lit.)

solubility

methanol: soluble

SMILES string

Sc1ccc(Cl)cc1

InChI

1S/C6H5ClS/c7-5-1-3-6(8)4-2-5/h1-4,8H

InChI key

VZXOZSQDJJNBRC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Chlorothiophenol undergoes oxidative coupling catalyzed by iron(III)-tetra phenyl prophyrin to form disulfides.

Application

4-Chlorothiophenol was used to modify poly(vinyl chloride) and to synthesize poly(vinyl chloride) with halogen groups.
Reactant involved in:
  • Oxidation to disulfides using cobalt-salen catalysts and air oxidizing agents
  • Thiourea-catalyzed sulfa-Michael addition to acryloyloxazolidinones
  • Pummerer-type cyclization
  • C-H Activation and C-S cross-coupling with heterocycles
  • Allylic sulfenylation with allylic carbonates
  • Lewis acid-catalyzed electrophilic ring-opening reactions of 2-aryl-3,4-dihydropyrans

related product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiongfeng Lin et al.
Nature communications, 8(1), 613-613 (2017-09-22)
Hybrid organic-inorganic halide perovskites are low-cost solution-processable solar cell materials with photovoltaic properties that rival those of crystalline silicon. The perovskite films are typically sandwiched between thin layers of hole and electron transport materials, which efficiently extract photogenerated charges. This
Selective Oxidation of Thiols to Disulfides Catalyzed by Iron (III)-Tetra Phenyl Porphyrin Using Urea-Hydrogen Peroxide as Oxidizing Reagent.
Karami B, et al.
Turkish Journal of Chemistry, 29(5), 539-539 (2005)
Xuejing Chen et al.
The Analyst, 144(14), 4312-4319 (2019-06-13)
A deep learning network called "residual neural network" (ResNet) was used to decode Raman spectra-encoded suspension arrays (SAs). With narrow bandwidths and stable signals, Raman spectra have ideal encoding properties. The different Raman reporter molecules assembled micro-quartz pieces (MQPs) were
Modification of poly (vinyl chloride) with new aromatic thiol compounds. Synthesis and characterization.
Navarro R, et al.
Polymer Degradation and Stability, 93(3), 585-591 (2008)
Homan Kang et al.
International journal of molecular sciences, 22(4) (2021-02-14)
We present a template-assisted method for synthesizing nanogap shell structures for biomolecular detections based on surface-enhanced Raman scattering. The interior nanogap-containing a silver shell structure, referred to as a silver nanogap shell (Ag NGS), was fabricated on silver nanoparticles (Ag

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service