Merck
  • Fabrication of graphene/gold-modified screen-printed electrode for detection of carcinoembryonic antigen.
Fabrication of graphene/gold-modified screen-printed electrode for detection of carcinoembryonic antigen.
Materials science & engineering. C, Materials for biological applications (2015-10-20)
K F Chan, H N Lim, N Shams, S Jayabal, A Pandikumar, N M Huang
ABSTRACT
Immunosensors based on gold nanoparticles and reduced graphene oxide (AuNPs/rGO)-modified screen-printed electrodes (SPEs) were successfully synthesized using an electrochemical deposition method. The modified SPEs were characterized using a field emission scanning electron microscope (FESEM) and Raman spectroscopy to analyze the morphology and composition of AuNPs and rGO. Both the FESEM and Raman spectroscopy revealed that the AuNPs were successfully anchored on the thin film of rGO deposited on the surface of the SPEs. Characterization with a ferri-ferrocyanide couple [Fe(CN)6(3-/4-)] showed that the electron transfer kinetic between the analyte and electrode was enhanced after the modification with the AuNPs/rGO composite on the electrode surface, in addition to increasing the effective surface area of the electrode. The modified SPE was immobilized with a sandwich type immunosensor to mimic the ELISA (enzyme-linked immunosorbent assay) immunoassay. The modified SPE that was fortified with the sandwich type immunosensor exhibited double electrochemical responses in the detection of carcinoembryonic antigen (CEA), with linear ranges of 0.5-50 ng/mL and 250-2000 ng/mL and limits of detection of 0.28 ng/mL and 181.5 ng/mL, respectively.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, BioReagent, suitable for electrophoresis, 98.0-102.0%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
di-Sodium hydrogen phosphate dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, ACS reagent, ≥98%
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
di-Sodium hydrogen phosphate dihydrate, meets analytical specification of Ph. Eur., BP, 98.5-101% (calc. to the dried substance)
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, puriss. p.a., ACS reagent, ≥99.0% (T)
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
di-Sodium hydrogen phosphate dihydrate, puriss. p.a., reag. Ph. Eur., 98.5-101.0% (calc. to the dried substance)
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, BioXtra, for molecular biology, ≥99.5% (T)
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%