Merck
  • Individualizing oral busulfan dose after using a test dose in patients undergoing hematopoietic stem cell transplantation: pharmacokinetic characterization.
Individualizing oral busulfan dose after using a test dose in patients undergoing hematopoietic stem cell transplantation: pharmacokinetic characterization.
Therapeutic drug monitoring (2015-01-15)
Cristiane Effting, Adriano de Moraes Arantes, Luciana V Queiroz Labre, Wilsione J Carneiro, Jerônimo R de Oliveira Neto, César Bariani, Caroline R Rodrigues, Andryne R Rodrigues, Luiz C Cunha
ABSTRACT
Busulfan is an alkylating agent used for conditioning patients undergoing hematopoietic stem cell transplantation with a narrow therapeutic range and highly variable pharmacokinetics. High concentrations induce toxicity, especially hepatic veno-occlusive disease, also referred to as sinusoidal obstruction syndrome. This study aimed to assess busulfan pharmacokinetic variability in pretransplant conditioning regimens using an analytical method validated by high-performance liquid chromatography coupled to diode array detector (HPLC/PDA). Eight patients who used the test dose (TD) of 1 mg/kg busulfan 10 days before conditioning were included, and 10 serial blood samples were collected to determine: the elimination half-life (t1/2), total area under the curve (AUCT), total clearance (Cl(T)/F), and plasma concentration at steady state (C(ss)), using a monocompartmental model and first-order kinetics. The instrumental conditions were: HPLC/PDA Shimadzu, column ACE C18 (150 mm × 4 mm); methanol/water/acetonitrile (65:20:15) eluent flow rate of 1 mL/min; 1,6-bis-(methanesulfonyloxy)-hexane; UV λ = 276 nm; analysis time 17 minutes; and derivatization with sodium diethylcarbamate. The dose was adjusted, and 4 blood samples per day were collected at days 2, 3, and 4 of treatment for new plasma determinations. Four patients needed higher doses; the mean dose administered was 1.02 ± 0.19 mg/kg. Mean results at TD: t1/2 = 2.88 ± 0.5 hours; Cl(T)/F = 0.18 ± 0.03 L · h(-1) · kg(-1); AUC(T) = 5461.00 ± 961.15 ng · mL(-1) · h(-1); and C(ss) = 911.3 ± 159.8 ng/mL. Mean results of samples collected during conditioning: t1/2 = 3.21 ± 0.9 hours; Cl(T)/F = 0.13 ± 0.02 L · h(-1) · kg(-1); AUC(T) = 7571 ± 1705 ng · mL(-1) · h(-1); and C(ss) = 1262.0 ± 284.3 ng/mL. High variability in the assessed pharmacokinetic parameters was observed, with a 38% variation in C(ss) between TD and conditioning regimen; Cl(T)/F decreased by 30%, suggesting drug accumulation after multiple-dose regimen. Although being lower than reported in the literature, this variation may be associated with toxicity of the proposed treatment, justifying patient monitoring and enhancing validity of previous pharmacokinetic evaluation using TD regimen.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Busulfan, analytical standard, for drug analysis
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Supelco
Hexane, analytical standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ultrapure Acetonitrile
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Busulfan, European Pharmacopoeia (EP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%