Skip to Content
Merck
All Photos(1)

Key Documents

B1170000

Busulfan

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Busulfan, 1,4-Butanediol dimethanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO2O(CH2)4OSO2CH3
CAS Number:
Molecular Weight:
246.30
Beilstein:
1791786
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

busulfan

manufacturer/tradename

EDQM

mp

114-117 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CS(=O)(=O)OCCCCOS(C)(=O)=O

InChI

1S/C6H14O6S2/c1-13(7,8)11-5-3-4-6-12-14(2,9)10/h3-6H2,1-2H3

InChI key

COVZYZSDYWQREU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Busulfan EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1A - Muta. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher Bredeson et al.
Blood, 122(24), 3871-3878 (2013-10-02)
We conducted a prospective cohort study testing the noninferiority of survival of ablative intravenous busulfan (IV-BU) vs ablative total body irradiation (TBI)-based regimens in myeloid malignancies. A total of 1483 patients undergoing transplantation for myeloid malignancies (IV-BU, N = 1025;
Je-Hwan Lee et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 31(6), 701-709 (2012-11-07)
We conducted a phase III randomized clinical trial to compare two myeloablative conditioning regimens for allogeneic hematopoietic cell transplantation (HCT) in patients with leukemia and myelodysplastic syndrome. After randomization, 64 patients received busulfan (3.2 mg/kg per day × 4 days)
Edward A Copelan et al.
Blood, 122(24), 3863-3870 (2013-09-26)
Cyclophosphamide combined with total body irradiation (Cy/TBI) or busulfan (BuCy) are the most widely used myeloablative conditioning regimens for allotransplants. Recent data regarding their comparative effectiveness are lacking. We analyzed data from the Center for International Blood and Marrow Transplant
J S McCune et al.
Clinical pharmacokinetics, 39(2), 155-165 (2000-09-08)
High dosage busulfan (1 mg/kg orally every 6 hours x 16 doses) is frequently used in preparative regimens for haemopoietic stem cell transplantation (HSCT). Busulfan is well absorbed after oral administration, exhibits low protein binding and is metabolised through conjugation
M Hassan et al.
Leukemia & lymphoma, 22(5-6), 395-407 (1996-08-01)
Busulfan as a high-dose therapy is an important component of many of the myeloablative regimens for both allogeneic and autologous bone marrow transplantation (BMT) in adults and children. During the last decade, several studies have shown a wide inter- and

Questions

  1. What's the pack size for this product?

    1 answer
    1. Each vial contains 50 mg.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service