Merck
  • Preparation of visible-light nano-photocatalysts through decoration of TiO2 by silver nanoparticles in inverse miniemulsions.
Preparation of visible-light nano-photocatalysts through decoration of TiO2 by silver nanoparticles in inverse miniemulsions.
Journal of colloid and interface science (2014-09-14)
Zhihai Cao, Shudi Zhu, Hui Qu, Dongming Qi, Ulrich Ziener, Liu Yang, Yingjie Yan, Haitang Yang
ABSTRACT
Ag/TiO2 nanocomposites were prepared through combination of a sol-gel process of a titanium precursor in inverse miniemulsions and in situ reduction of silver ions in the "nanoreactors". The morphological investigation shows that Ag nanoparticles are mainly located on the surface of TiO2 nano-supports because of the fast reduction rate of Ag ions by hydrazine. Ag/TiO2 nanocomposites with amorphous or anatase TiO2 phase displayed high visible-light catalytic activity for degradation of Rhodamine B. The photoactivity of Ag/anatase TiO2 nanocomposites could be influenced by the Ag content that could be conveniently tuned by the loading of silver salts. The influence of the loading of silver salts on the particle properties of the Ag/TiO2 nanocomposites was investigated systematically.
Sigma-Aldrich
Titanium(IV) ethoxide
Sigma-Aldrich
Rhodamine B, Dye content 90 %
Sigma-Aldrich
Titanium(IV) ethoxide, technical grade
Supelco
Rhodamine B solution, 0.2% in isopropanol, for TLC derivatization
Sigma-Aldrich
Rhodamine B, ≥95% (HPLC)
Supelco
Rhodamine B, analytical standard
Sigma-Aldrich
Rhodamine B, for fluorescence
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard