Merck
  • Neonatal outcomes after the implantation of human embryos vitrified using a closed-system device.
Neonatal outcomes after the implantation of human embryos vitrified using a closed-system device.
Journal of assisted reproduction and genetics (2015-01-27)
Hideyuki Iwahata, Shu Hashimoto, Masayasu Inoue, Tomoko Inoue, Keijiro Ito, Yoshiharu Nakaoka, Nao Suzuki, Yoshiharu Morimoto
ABSTRACT
Closed vitrification poses a risk of adversely affecting embryo development, while it may minimize the risk of contamination. We assessed the effects of closed-system human embryo vitrification on fetal development after implantation, neonatal outcome, and clinical safety. This was a retrospective cohort study conducted at a private fertility clinic. A total of 875 vitrified-warmed blastocysts that were single-transferred under hormone-replacement cycles between November 2011 and December 2013 were randomly divided into two groups (closed vitrification, n 313; open vitrification, n 562) after receiving the patients' consent forms. Developmental competence after implantation, including gestational age, birth weight, sex, Apgar score, and anomalies of newborns, after the transfer of blastocysts vitrified by closing vitrification was compared with that obtained in the case of open vitrification. There were no significant differences between the use of closed and open vitrification systems in embryo development after implantation, gestational age, birth weight, sex ratio, Apgar score, and congenital anomalies of newborns. Human embryos can be vitrified using a closed vitrification system without impairment of neonatal development.
Product Description
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
Progesterone, European Pharmacopoeia (EP) Reference Standard
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Luteinizing hormone releasing hormone salmon, ≥97% (HPLC)
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Progesterone, ≥99%
Supelco
Progesterone, VETRANAL®, analytical standard
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Sucrose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylene glycol 5 M solution
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Estradiol valerate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF