Skip to Content
Merck
All Photos(3)

Key Documents

324558

Sigma-Aldrich

Ethylene glycol

anhydrous, 99.8%

Synonym(s):

1,2-Ethanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2OH
CAS Number:
Molecular Weight:
62.07
Beilstein:
505945
EC Number:
MDL number:
UNSPSC Code:
12191502
eCl@ss:
39020501
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
bp:
195-198 °C
vapor pressure:
0.08 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

vapor density

2.1 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)

Assay

99.8%

form

liquid

autoignition temp.

752 °F

expl. lim.

15.3 %

impurities

<0.003% water
<0.005% water (100 mL pkg)

refractive index

n20/D 1.431 (lit.)

bp

195-198 °C

mp

−13 °C (lit.)

density

1.113 g/mL at 25 °C (lit.)

SMILES string

OCCO

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethylene glycol is the major component of antifreeze preparations.[1]

Application

Ethylene glycol may be used in the preparation of spiroborate esters by reacting with chiral 1,2-aminoalcohols in the presence of triisopropyl borate.[2] It may also be used as a solvent in the preparation of tris(2,2′-bipyrazyl)ruthenium(II) chloride by the reaction of 2,2′-bipyrazine with ruthenium(III) chloride.[3]

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

239.0 °F - open cup

Flash Point(C)

115 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bis(tetrakis(3,5-bis(trifluoromethyl)phenyl)borate)
Lies SD, et al.
Organic Syntheses, 93, 178-178 (2016)
The spectrum of ethylene glycol poisoning.
Moriarty RW & McDonald RH.
Clinical Toxicology, 7(6), 583-596 (1974)
Synthesis of spiroborate esters from 1,2-aminoalcohols, ethylene glycol and triisopropyl borate: preparation of (S)-1-(1,3,2-dioxaborolan-2-yloxy)-3-methyl-1,1-diphenylbutan-2-amine
Stepanenko V, et al.
Organic Syntheses, 87, 26-26 (2010)
Gregory Charles Nordmann et al.
Scientific reports, 10(1), 915-915 (2020-01-24)
The transcription factor ZENK is an immediate early gene that has been employed as a surrogate marker to map neuronal activity in the brain. It has been used in a wide variety of species, however, commercially available antibodies have limited
Birgit Oberleitner et al.
Chemical communications (Cambridge, England), 49(16), 1615-1617 (2013-01-23)
This work describes an integrated approach for designing on demand Self-Assembled Monolayers (SAMs) on silicon oxides and particularly glass substrates for cell biology applications. Starting from commercially available compounds, the strategy relies on thiol-ene reaction and provides high quality SAMs

Questions

1–5 of 5 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. what is the expiry data of Ethylene glycol anhydrous 99.8% reagent of Batch SHBL6653 with product code 324558.

    1 answer
    1. This product is not assigned an expiration date or recommended retest date. Customers should routinely inspect products with no expiration date or recommended retest date to ensure they perform as expected. These products are also subject to a one-year warranty from the date of shipment.

      For more information, you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

  4. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  5. What is the molarity of ethylene glycol?

    1 answer
    1. Ethylene glycol is 17.93M.  This is calculated using the density (1.113 g/ml) and the molecular weight (62.07 g/mol).  If the product is less than 100% pure, correct for the purity.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service