Skip to Content
Merck
All Photos(1)

Key Documents

1250008

USP

Estradiol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

β-Estradiol, 1,3,5-Estratriene-3,17β-diol, 17β-Estradiol, 3,17β-Dihydroxy-1,3,5(10)-estratriene, Dihydrofolliculin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24O2
CAS Number:
Molecular Weight:
272.38
Beilstein:
1914275
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

estradiol

manufacturer/tradename

USP

mp

176-180 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O[C@H]1CC[C@@]2([H])[C@]3([H])CCC4=CC(O)=CC=C4[C@@]3([H])CC[C@@]21C

InChI

1S/C18H24O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-17,19-20H,2,4,6-9H2,1H3/t14-,15-,16+,17+,18+/m1/s1

InChI key

VOXZDWNPVJITMN-ZBRFXRBCSA-N

Gene Information

human ... ESR1(2099)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

The major estrogen secreted by the premenopausal ovary.
The major estrogen secreted by the premenopausal ovary. Estrogens direct the development of the female phenotype in embryogenesis and during puberty by regulating gene transcription and, thus, protein synthesis. It also induces the production of gonadotropins which, in turn, induce ovulation. Exposure to estradiol increases breast cancer incidence and proliferation.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Estradiol and Norethindrone Acetate Tablets
United States Pharmacopeia, 44(6) (2023)
Estradiol Cypionate
United States Pharmacopeia, 39(6), 1737-1737 (2020)
Marie Mi Bonde et al.
Nucleic acids research, 42(20), 12847-12860 (2014-10-30)
Splicing reactions generally combine high speed with accuracy. However, some of the pre-mRNAs escape the nucleus with a retained intron. Intron retention can control gene expression and increase proteome diversity. We calculated the escape rate for the yeast PTC7 intron
Amit Cohen et al.
Zebrafish, 11(5), 462-478 (2013-06-19)
Estrogen is a steroid hormone that has been implicated in a variety of cellular and physiological processes and in the development of diseases such as cancer. Here we show a remarkable widespread microRNA (miRNA) downregulation in the zebrafish (Danio rerio)
Mark A Tarnopolsky
Medicine and science in sports and exercise, 40(4), 648-654 (2008-03-05)
Women oxidize more lipid and less carbohydrate and protein compared with men during endurance exercise. The increase in fat oxidation is associated with higher intramyocellular lipid content and use as well as greater adipocyte lipolysis. Glucose rates of appearance and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service