Merck
  • Human B cells induce dendritic cell maturation and favour Th2 polarization by inducing OX-40 ligand.
Human B cells induce dendritic cell maturation and favour Th2 polarization by inducing OX-40 ligand.
Nature communications (2014-06-10)
Mohan S Maddur, Meenu Sharma, Pushpa Hegde, Emmanuel Stephen-Victor, Bali Pulendran, Srini V Kaveri, Jagadeesh Bayry
ABSTRACT
Dendritic cells (DCs) play a critical role in immune homeostasis by regulating the functions of various immune cells, including T and B cells. Notably, DCs also undergo education on reciprocal signalling by these immune cells and environmental factors. Various reports demonstrated that B cells have profound regulatory functions, although only few reports have explored the regulation of human DCs by B cells. Here we demonstrate that activated but not resting B cells induce maturation of DCs with distinct features to polarize Th2 cells that secrete interleukin (IL)-5, IL-4 and IL-13. B-cell-induced maturation of DCs is contact dependent and implicates signalling of B-cell activation molecules CD69, B-cell-activating factor receptor, and transmembrane activator and calcium-modulating cyclophilin ligand interactor. Mechanistically, differentiation of Th2 cells by B-cell-matured DCs is dependent on OX-40 ligand. Collectively, our results suggest that B cells have the ability to control their own effector functions by enhancing the ability of human DCs to mediate Th2 differentiation.
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, meets USP testing specifications
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, ≥98% (HPLC), γ-irradiated, suitable for cell culture
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Human IL21 / Interleukin-21 ELISA Kit
Sigma-Aldrich
TSLP human, recombinant, expressed in E. coli, ≥97% (SDS-PAGE), ≥97% (HPLC)
Sigma-Aldrich
Mouse IL-21 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Human CXCL16 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Mouse CXCL16 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Human IL-22 ELISA Kit, for serum, plasma, cell culture supernatant and urine