Merck
  • Exploring metabolic syndrome serum free fatty acid profiles based on GC-SIM-MS combined with random forests and canonical correlation analysis.
Exploring metabolic syndrome serum free fatty acid profiles based on GC-SIM-MS combined with random forests and canonical correlation analysis.
Talanta (2015-02-03)
Ling Dai, Carlos M Vicente Gonçalves, Zhang Lin, Jianhua Huang, Hongmei Lu, Lunzhao Yi, Yizeng Liang, Dongsheng Wang, Dong An
ABSTRACT
Metabolic syndrome (MetS) is a cluster of metabolic abnormalities associated with an increased risk of developing cardiovascular diseases or type II diabetes. Till now, the etiology of MetS is complex and still unknown. Metabolic profiling is a powerful tool for exploring metabolic perturbations and potential biomarkers, thus may shed light on the pathophysiological mechanism of diseases. In this study, fatty acid profiling was employed to exploit the metabolic disturbances and discover potential biomarkers of MetS. Fatty acid profiles of serum samples from metabolic syndrome patients and healthy controls were first analyzed by gas chromatography-selected ion monitoring-mass spectrometry (GC-SIM-MS), a robust method for quantitation of fatty acids. Then, the supervised multivariate statistical method of random forests (RF) was used to establish a classification and prediction model for MetS, which could assist the diagnosis of MetS. Furthermore, canonical correlation analysis (CCA) was employed to investigate the relationships between free fatty acids (FFAs) and clinical parameters. As a result, several FFAs, including C16:1n-9c, C20:1n-9c and C22:4n-6c, were identified as potential biomarkers of MetS. The results also indicated that high density lipoprotein-cholesterol (HDL-C), triglycerides (TG) and fasting blood glucose (FBG) were the most important parameters which were closely correlated with FFAs disturbances of MetS, thus they should be paid more attention in clinical practice for monitoring FFAs disturbances of MetS than waist circumference (WC) and systolic blood pressure/diastolic blood pressure (SBP/DBP). The results have demonstrated that metabolic profiling by GC-SIM-MS combined with RF and CCA may be a useful tool for discovering the perturbations of serum FFAs and possible biomarkers for MetS.
Product Description
Supelco
Methanol, analytical standard
Supelco
Sulfuric acid, for the determination of nitrogen, for inorganic trace analysis, ≥97.0%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
cis-10-Nonadecenoic acid, ≥99%, liquid
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard