Skip to Content
Merck
All Photos(1)

Documents

84736

Sigma-Aldrich

Sulfuric acid solution

puriss. p.a., ≥25% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

grade

puriss. p.a.

Quality Level

form

liquid

concentration

≥25% (T)

impurities

≤0.0008% KMnO4-reducing matter (as O)

ign. residue

≤0.0005%

density

1.80-1.84 g/cm3

anion traces

chloride (Cl-): ≤0.5 mg/kg
nitrate (NO3-): ≤0.2 mg/kg

cation traces

Ag: ≤0.02 mg/kg
Al: ≤0.05 mg/kg
As: ≤0.01 mg/kg
Ba: ≤0.05 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.2 mg/kg
Cd: ≤0.02 mg/kg
Co: ≤0.01 mg/kg
Cr: ≤0.05 mg/kg
Cu: ≤0.01 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.1 mg/kg
Li: ≤0.01 mg/kg
Mg: ≤0.05 mg/kg
Mn: ≤0.01 mg/kg
Mo: ≤0.05 mg/kg
NH4+: ≤2 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.02 mg/kg
Sr: ≤0.01 mg/kg
Tl: ≤0.05 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulfuric acid is a strong mineral acid. It participates as a medium in the anodic oxidation of aniline at a platinum electrode. Anodic oxidation has been reported to proceed via a free radical mechanism and octamer emeraldimine has been obtained as a major product.

Application

Sulfuric acid solution has been used as a solvent for the preparation of 1M xylenol orange and 2.5mM ammonium ferrous sulfate solution in a study.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Omidreza Firuzi et al.
Journal of clinical laboratory analysis, 20(4), 139-148 (2006-07-29)
It has been proposed that sample storage may have some influence on the parameters of oxidative stress status (OSS) in biological fluids. We measured four important OSS parameters in plasma of 23 healthy subjects and repeated the measurements in the
Investigation of the kinetics and mechanism of the anodic oxidation of aniline in aqueous sulfuric acid solution at a platinum electrode.
Mohilner DM, et al.
Journal of the American Chemical Society, 84(19), 3618-3622 (1962)
I A Panagiotopoulos et al.
Bioresource technology, 130, 570-577 (2013-01-22)
A two-stage pretreatment approach, employing steam followed by organosolv treatment, was assessed for its ability to fractionate and recover most of the hemicellulose, lignin and cellulose components of poplar wood chips. A mild steaming stage was initially used to maximise
João Almeida et al.
Nature, 502(7471), 359-363 (2013-10-08)
Nucleation of aerosol particles from trace atmospheric vapours is thought to provide up to half of global cloud condensation nuclei. Aerosols can cause a net cooling of climate by scattering sunlight and by leading to smaller but more numerous cloud
Francesco Riccobono et al.
Science (New York, N.Y.), 344(6185), 717-721 (2014-05-17)
Atmospheric new-particle formation affects climate and is one of the least understood atmospheric aerosol processes. The complexity and variability of the atmosphere has hindered elucidation of the fundamental mechanism of new-particle formation from gaseous precursors. We show, in experiments performed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service