Skip to Content
Merck
All Photos(1)

Documents

439193

Sigma-Aldrich

Methanol

≥99.9%, gradient grade, suitable for HPLC, suitable for EPA 1613

Synonym(s):

Methyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein:
1098229
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.05

product name

Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%

grade

gradient grade

Quality Level

Agency

suitable for EPA 1613

vapor density

1.11 (vs air)

vapor pressure

410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

Assay

≥99.9%

form

liquid

autoignition temp.

725 °F

expl. lim.

36 %

technique(s)

HPLC: suitable

impurities

H2SO4, passes test (darkened)
≤0.0002 meq/g Titr. base
≤0.0003 meq/g Titr. acid
≤0.001% acetaldehyde
≤0.001% acetone
≤0.001% formaldehyde
≤0.03% water

evapn. residue

≤0.0003%

color

APHA: ≤10
clear

refractive index

n20/D 1.329 (lit.)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

HPLC-gradient

≤5 mAU at 254 nm

λ

H2O reference

UV absorption

λ: 203 nm Amax: ≤1.0
λ: 210 nm Amax: ≤0.60
λ: 220 nm Amax: ≤0.30
λ: 230 nm Amax: ≤0.15
λ: 240 nm Amax: ≤0.05
λ: 260-400 nm Amax: ≤0.01

suitability

passes test for HPLC

application(s)

food and beverages

format

neat

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methanol (MeOH) is an organic solvent with a wide range of industrial applications. It is the main component of lithium batteries. It can be prepared by hydrogenation of CO2 under various conditions. MeOH is a toxic alcohol employed for denaturing ethyl alcohol. A study reports the preparation of biodiesel from canola oil using methanol under supercritical conditions.

Application

High-purity, multipurpose solvent, tested for suitability in:
  • HPLC with gradient analysis
  • Spectrophotometry
  • Residual testing
  • LC-MS applications

Used as a solvent/reagent to determine total phenols content, anti-DPPH activity and the content of antioxidants in honeybee drone blood homogenate (DBH) samples.
Meets ACS specifications.

Preparation Note

Product filtered through a 0.2 μm filter

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Combining Low-Pressure CO2 Capture and Hydrogenation To Form Methanol.
Khusnutdinova JR, et al.
ACS Catalysis, 5(4), 2416-2422 (2015)
R A Cunha et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 18(6), 1987-1995 (1998-05-15)
ATP analogs substituted in the gamma-phosphorus (ATPgammaS, beta, gamma-imido-ATP, and beta,gamma-methylene-ATP) were used to probe the involvement of P2 receptors in the modulation of synaptic transmission in the hippocampus, because their extracellular catabolism was virtually not detected in CA1 slices.
Ufuk Cobanoglu et al.
Asian Pacific journal of cancer prevention : APJCP, 12(6), 1399-1403 (2011-12-01)
Early diagnosis and prevention is very important for lung cancer patients. Previous studies have emphasized that the level of coenzyme Q10 (CoQ10), present primarily in mitochondria, decreases with age and is low in patients with chronic diseases. Our goal was
Nomaan M Rezayee et al.
Journal of the American Chemical Society, 137(3), 1028-1031 (2015-01-17)
This Communication describes the hydrogenation of carbon dioxide to methanol via tandem catalysis with dimethylamine and a homogeneous ruthenium complex. Unlike previous examples with homogeneous catalysts, this CO2-to-CH3OH process proceeds under basic reaction conditions. The dimethylamine is proposed to play
Methanol.
R Von Burg
Journal of applied toxicology : JAT, 14(4), 309-313 (1994-07-01)

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service