Skip to Content
Merck
All Photos(2)

Documents

A6390

Sigma-Aldrich

β-D-Allose

rare aldohexose sugar

Synonym(s):

β-D-Allopyranose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)
≥98% (TLC)

form

powder

optical activity

[α]/D 12.00 to 16.00 °, c = 45.00-55.00 mg/mL in water

technique(s)

HPLC: suitable
thin layer chromatography (TLC): suitable

impurities

<2.00% water (Karl Fischer)

color

white to yellow cast

solubility

water: 49.00-51.00 mg/mL, clear, colorless to light yellow

SMILES string

OC[C@H]1O[C@@H](O)[C@H](O)[C@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4-,5-,6-/m1/s1

InChI key

WQZGKKKJIJFFOK-QZABAPFNSA-N

Application

β-D-Allose (β-D-Allose), a rare sugar member of the aldohexose family and a C3 epimer of glucose, is used as an inhibitor of fruiting body formation and sporulation in Myxococcus xanthus.

Biochem/physiol Actions

D-Allose is a rare aldohexose sugar that has demonstrated anti-cancer, antitumor, anti-inflammatory, anti-oxidative, antihypertensive, cryoprotective, and immunosuppressant activities. D-Allose upregulates thioredoxin-interacting protein (TXNIP) gene expression and was found to induce G1 cell cycle arrest in hepatocellular carcinoma cells. It inhibited transcriptional activity of the activator protein 1, nuclear factor-κB, and nuclear factor of activated T cells. D-Allose decreases transport of 2-deoxy-D-glucose and 3-O-methyl-D-glucose in V79 Chinese hamster lung fibroblast cells.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R J Germinario et al.
Journal of cellular physiology, 145(2), 318-323 (1990-11-01)
Exposure to D-allose has been demonstrated to lead to decreased 2-deoxy-D-glucose (2-DG) and 3-0-methyl-D-glucose transport in the V79 Chinese hamster lung fibroblast cell line. The effect of D-allose 1) was maximal after 4 hours exposure to the cells; 2) was
Kana Yamada et al.
Nutrition research (New York, N.Y.), 32(2), 116-123 (2012-02-22)
Oxidative stress modulates the osteoclast differentiation via redox systems, and thioredoxin 1 (Trx) promotes the osteoclast formation by regulating the activity of transcription factors. The function of Trx is known to be regulated by its binding partner, thioredoxin-interacting protein (TXNIP).
Fuminori Yamaguchi et al.
International journal of oncology, 32(2), 377-385 (2008-01-19)
'Rare sugars' are defined as monosaccharides that exist in nature but are only present in limited quantities. The development of mass production method of rare sugars revealed some interesting physiological effects of these on animal cells, but the mechanisms have
Aditya Venkatesh et al.
The Journal of clinical investigation, 125(4), 1446-1458 (2015-03-24)
Retinitis pigmentosa (RP) is an inherited photoreceptor degenerative disorder that results in blindness. The disease is often caused by mutations in genes that are specific to rod photoreceptors; however, blindness results from the secondary loss of cones by a still
Yu-Ri Lim et al.
Applied microbiology and biotechnology, 91(2), 229-235 (2011-06-10)
D-allose has attracted a great deal of attention in recent years due to its many pharmaceutical activities, which include anti-cancer, anti-tumor, anti-inflammatory, anti-oxidative, anti-hypertensive, cryoprotective, and immunosuppressant activities. D-allose has been produced from D-psicose using D-allose-producing enzymes, including L-rhamnose isomerase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service