Merck
  • Simultaneous determination of 38 veterinary antibiotic residues in raw milk by UPLC-MS/MS.
Simultaneous determination of 38 veterinary antibiotic residues in raw milk by UPLC-MS/MS.
Food chemistry (2015-03-22)
R W Han, N Zheng, Z N Yu, J Wang, X M Xu, X Y Qu, S L Li, Y D Zhang, J Q Wang
ABSTRACT
A selective and rapid method has been developed to determine, simultaneously, 38 veterinary antibiotic residues in raw milk by ultra-high-performance liquid chromatography-tandem mass spectrometry (UPLC-MS/MS). One milliliter of raw milk was diluted with 0.5 mL water and 3 mL acetonitrile, then purified using an Oasis HLB cartridge. The eluates were evaporated by nitrogen drying and then reconstituted to 4 mL with water/acetonitrile (8:1) before being injected into the UPLC-MS/MS system. The results indicated recoveries of 68-118% for 14 β-lactams, 79-118% for eight quinolones, 71-106% for eight sulfonamides, 76-116% for four tetracyclines, 78-106% for three macrolides, and 88-103% for one lincosamides, with coefficients of variation less than 15% for intraday and interday precisions. The limit of quantification for all antibiotics was 0.03-10 μg kg(-1). This methodology was then applied to field-collected real raw milk samples and trace levels of four antibiotics were detected.
Product Description
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Supelco
Sulfamethoxazole, VETRANAL®, analytical standard
Supelco
Sulfapyridine, VETRANAL®, analytical standard
Supelco
Enrofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Sulfamethoxazole, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ciprofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sulfamethoxazole, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Enrofloxacin, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic, purum p.a., anhydrous, ≥98.0% (T)
Supelco
Sulfachloropyridazine, VETRANAL®, analytical standard
Supelco
Sulfadiazine, VETRANAL®, analytical standard
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Supelco
Sulfapyridine, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%