Skip to Content
Merck
All Photos(3)

Key Documents

31573

Supelco

5α-Androstan-17β-ol-3-one

VETRANAL®, analytical standard

Synonym(s):

17β-Hydroxy-5α-androstan-3-one, 4,5α-Dihydrotestosterone, Androstanolone, Stanolone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H30O2
CAS Number:
Molecular Weight:
290.44
Beilstein:
2056371
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

drug control

USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]12CC[C@@]3([H])[C@]4([H])CC[C@H](O)[C@@]4(C)CC[C@]3([H])[C@@]1(C)CCC(=O)C2

InChI

1S/C19H30O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h12,14-17,21H,3-11H2,1-2H3/t12-,14-,15-,16-,17-,18-,19-/m0/s1

InChI key

NVKAWKQGWWIWPM-ABEVXSGRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bu B Yeap et al.
The Journal of clinical endocrinology and metabolism, 99(1), E9-18 (2013-11-22)
Testosterone (T) levels decline with age and lower T has been associated with increased mortality in aging men. However, the associations of its metabolites, dihydrotestosterone (DHT) and estradiol (E2), with mortality are poorly defined. We assessed associations of T, DHT
Katharine Trenholme et al.
Antimicrobial agents and chemotherapy, 58(7), 3666-3678 (2014-04-16)
Therapies to prevent transmission of malaria parasites to the mosquito vector are a vital part of the global malaria elimination agenda. Primaquine is currently the only drug with such activity; however, its use is limited by side effects. The development
Takashi Taniguchi et al.
Arthritis & rheumatology (Hoboken, N.J.), 67(2), 517-526 (2014-11-12)
Fli-1, a potential predisposing factor for systemic sclerosis (SSc), is constitutively down-regulated in the lesional skin of patients with SSc by an epigenetic mechanism. To investigate the impact of Fli-1 deficiency on the induction of an SSc phenotype in various
Isabelle Brunet et al.
The Journal of clinical investigation, 124(7), 3230-3240 (2014-06-18)
Autonomic sympathetic nerves innervate peripheral resistance arteries, thereby regulating vascular tone and controlling blood supply to organs. Despite the fundamental importance of blood flow control, how sympathetic arterial innervation develops remains largely unknown. Here, we identified the axon guidance cue
Jennifer Downey et al.
Bone, 71, 164-170 (2014-12-03)
Skeletal muscle has strong regenerative capabilities. However, failed regeneration can lead to complications where aberrant tissue forms as is the case with heterotopic ossification (HO), in which chondrocytes, osteoblasts and white and brown adipocytes can arise following severe trauma. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service