Merck
  • An investigation into the drug release from ibuprofen matrix tablets with ethylcellulose and some poly-acrylate polymers.
An investigation into the drug release from ibuprofen matrix tablets with ethylcellulose and some poly-acrylate polymers.
Pakistan journal of pharmaceutical sciences (2014-05-09)
Hosseinali Tabandeh, Seyed Alireza Mortazavi
ABSTRACT
This study was performed to achieve sustained-release Ibuprofen matrix tablets with a zero-order release kinetic while most of the previous formulations have shown Higuchi release kinetic. Considering the results from previous studies, ethyl cellulose, Carbopol 934P, Carbopol 974P, and Pemulen TR-1 were used at different amounts for preparation of the tablets by direct compression. The release profiles were studied in a two-stage release test using non-linear regression analysis. Carbopols 934P and 974P could not sustain the release adequately while Pemulen TR-1 had too strong sustaining effect. Therefore, combination formulations were considered and studied. The release profiles of ethyl cellulose formulation and the combination formulation consisting Carbopol 934P and Pemulen TR-1 best fitted in Higuchi model, although the zero-order model was not completely rejected. However, the kinetic model of release from the combination formulation consisting Carbopol 974P and Pemulen TR-1 changed to zero-order indicating the most constant release rate among formulations. This was speculated to be due to some erosion of the gel, as well as some interaction of the hydrophobic chain of Pemulen TR-1 with Ibuprofen. Therefore, this formulation is suggested for directly compressed sustained-release matrix tablets of Ibuprofen with a more constant release rate.
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Supelco
Ibuprofen sodium salt, ≥98% (GC)
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
α-Cellulose, powder
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Ibuprofen, United States Pharmacopeia (USP) Reference Standard
Supelco
Avicel® PH-101, ~50 μm particle size
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ibuprofen
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
Supelco
Cellulose, powder, for column chromatography
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Supelco
Cellulose, DS-0, powder, suitable for thin layer chromatography (TLC)
Sigma-Aldrich
Ethyl cellulose, 48.0-49.5% (w/w) ethoxyl basis
Sigma-Aldrich
Cellulose, microcrystalline, powder
Supelco
Cellulose, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Sigma-Aldrich
Cellulose, colloidal, microcrystalline
Sigma-Aldrich
Ethyl cellulose, viscosity 46 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Ethyl cellulose, viscosity 22 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Ethyl cellulose, viscosity 4 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Ethyl cellulose, viscosity 10 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Ethyl cellulose, viscosity 300 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
(S)-(+)-Ibuprofen, ReagentPlus®, 99%
Sigma-Aldrich
Ethyl cellulose, viscosity 100 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm
Supelco
Cellulose, acid washed, powder, for column chromatography