Skip to Content
Merck
All Photos(3)

Key Documents

200697

Sigma-Aldrich

Ethyl cellulose

viscosity 22 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl

Synonym(s):

Ethylcellulose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

Quality Level

autoignition temp.

698 °F

concentration

47.5-49.5%

extent of labeling

48% ethoxyl

refractive index

n20/D 1.47 (lit.)

viscosity

22 cP, 5 % in toluene/ethanol 80:20(lit.)

transition temp

softening point 155 °C

density

1.14 g/mL at 25 °C (lit.)

SMILES string

O1[C@H](C(C(C(C1COCC)OCC)OCC)OCC)O[C@@H]2C(OC(C(C2OCC)OCC)OCC)COCC

InChI key

MINRZDZESGXKDM-OLIDUAMRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl cellulose is a non-toxic polysaccharide type gelator widely used in surface coatings and biomedical applications.

Application

Ethyl cellulose can be used:
  • To prepare oleogels for the ophthalmic delivery of hydrophilic drugs.
  • As a coating agent to synthesize flexible bioplastics for packaging applications.
  • To prepare CuO/Cu2O granules for thermochemical heat storage applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetsuya Ozeki et al.
Biological & pharmaceutical bulletin, 35(11), 1926-1931 (2012-11-06)
Production of drug nanoparticles is an effective strategy to enhance solubility and oral absorption of water-insoluble drugs. The handling of drug nanoparticles has been an important issue in drug formulation because nanoparticles easily aggregate each other and redispersion of these
H V Adikane et al.
Applied biochemistry and biotechnology, 169(3), 1026-1038 (2013-01-09)
The chemical modification of developed ethyl cellulose-based membrane was carried out to make it suitable for bioseparation. The different reagents were used for the modification of membrane to couple protein A (PA) to study the purification of immunoglobulin G (IgG)
Kifayat Ullah Shah et al.
TheScientificWorldJournal, 2012, 842348-842348 (2012-06-01)
The design and fabrication of sustained/controlled release dosage forms, employing new excipients capable of extending/controlling the release of drugs from the dosage forms over prolonged periods, has worked well in achieving optimally enhanced therapeutic levels of the drugs. In this
D G Yu et al.
Acta biomaterialia, 9(3), 5665-5672 (2012-10-27)
The capability of core/sheath nanofibers prepared using coaxial electrospinning to provide adjustable biphasic drug release was investigated. Using ketoprofen (KET) as the model drug, polyvinylpyrrolidone as the sheath polymer, and ethyl cellulose as the core matrix, the coaxial process could
Anurag Verma et al.
Acta pharmaceutica (Zagreb, Croatia), 62(2), 237-250 (2012-07-04)
Chitosan has become a focus of major interest in recent years due to its excellent biocompatibility, biodegradability and non-toxicity. Although this material has already been extensively investigated in the design of different types of drug delivery systems, it is still

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service