Merck
  • [The prevention of the development of the collagen-induced microembolic syndrome by decreasing the hydrodynamic resistance of the blood with special polymers under flow].
[The prevention of the development of the collagen-induced microembolic syndrome by decreasing the hydrodynamic resistance of the blood with special polymers under flow].
Patologicheskaia fiziologiia i eksperimental'naia terapiia (1991-07-01)
I L Konorova, S E Akopov, E S Gabrielian, I V Gannushkina
ABSTRACT
The search for antiaggregatory compounds is undertaken, as a rule, under in vitro conditions which do not reflect the dynamics of the real process. The present work deals with study of the peculiarities of the development of the collagen induced microembolic syndrome and the effect produced on it by hydrodynamic resistance reducing polymers under conditions modelling real circulation. The authors show the formation of a developed microembolic syndrome under the effect of collagen with accumulation of vasoconstrictors in the environment and increase of red cell hemolysis. Reduction of blood flow turbulence by the addition of solutions of neutral high-molecular linear polymers revealed the inhibiting effect of these polymers on the development of intracellular aggregate formation and concomitant red cell microhemolysis, which was attended by sharp reduction of thromboxane A2 production in the experiment. The authors show the significance of hemodynamic conditions in the development of the microembolic syndrome and the possibility of preventing it under conditions of modification of the blood flow with polymers, which may be of high prophylactic importance.
Sigma-Aldrich
Polyethylene glycol solution, BioUltra, for molecular biology, 1,000, ~50% in H2O
Sigma-Aldrich
Poly(ethylene glycol), average Mn 3,350, powder
Sigma-Aldrich
Poly(ethylene glycol), average Mv ~8,000, powder (crystalline)
Sigma-Aldrich
Poly(ethylene glycol), average Mn 600, waxy solid (moist)
Supelco
Poly(ethylene glycol), analytical standard, for GPC, 40,000
Supelco
Poly(ethylene glycol), analytical standard, for GPC, 1,000
Sigma-Aldrich
Poly(ethylene glycol), average mol wt 200
Sigma-Aldrich
Poly(ethylene glycol), average mol wt 1,450
Sigma-Aldrich
Poly(ethylene glycol), average mol wt 400
Sigma-Aldrich
Poly(ethylene glycol), BioXtra, average mol wt 3,350, powder
Sigma-Aldrich
Poly(ethylene glycol), average mol wt 8,000, powder
Sigma-Aldrich
Poly(ethylene glycol), average Mn 950-1,050
Sigma-Aldrich
Poly(ethylene glycol), for molecular biology, average mol wt 8,000
Sigma-Aldrich
Poly(ethylene glycol), Hybri-Max, mol wt 1,300-1,600, waxy solid, BioReagent, suitable for hybridoma
Sigma-Aldrich
Polyethylene glycol solution, 40 % (w/w) in H2O, average mol wt 8,000
Supelco
Poly(ethylene glycol), analytical standard, for GPC, 20,000
Sigma-Aldrich
Poly(ethylene glycol), tested according to Ph. Eur., 400
Sigma-Aldrich
Poly(ethylene oxide), average Mv 400,000 (nominal), powder
Sigma-Aldrich
Poly(ethylene oxide), average Mv 100,000 (nominal), powder
Sigma-Aldrich
Poly(ethylene oxide), average Mv ~4,000,000 (nominal), contains <1000 ppm BHT as inhibitor
Sigma-Aldrich
Poly(ethylene oxide), average Mv ~5,000,000 (nominal), powder
Sigma-Aldrich
Poly(ethylene oxide), average Mv 200,000 (nominal), powder
Sigma-Aldrich
Poly(ethylene oxide), average Mv ~900,000 (nominal), powder
Sigma-Aldrich
Poly(ethylene glycol), average Mn 2,050, chips
Sigma-Aldrich
Poly(ethylene oxide), average Mv ~2,000,000 (nominal), powder
Sigma-Aldrich
Poly(ethylene oxide), average Mv ~8,000,000 (nominal), powder
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 4,000
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 3,350
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 400
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 20,000