Skip to Content
Merck
All Photos(1)

Key Documents

81310

Sigma-Aldrich

Poly(ethylene glycol)

average MN 35,000, hydroxyl

Synonym(s):

Polyethylene glycol, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.23

Product Name

Poly(ethylene glycol), 35,000

form

flakes

mol wt

average Mn 35,000

mp

64-66 °C

Ω-end

hydroxyl

α-end

hydroxyl

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) (PEG) is a synthetic polymer that is an oligomer of ethylene oxide with properties such as cost efficiency, hydrophilicity, and biocompatibility that make it suitable in the production of biomedical hydrogels.

Application

PEG can be used for a wide range of applications in the field of drug delivery, biomedical applications and energy storage systems.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

281.5 °F - closed cup

Flash Point(C)

138.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrogels in craniofacial tissue engineering
Biomaterials for Oral and Dental Tissue Engineering, 47-64 (2017)
Entrapment of essential oils in hydrogels for biomedical applications
Polymeric Gels, 125-141 (2018)
Biomedical hydrogels
Biomaterials, Artificial Organs and Tissue Engineering, 107-115 (2005)
Oğuz Kaan Kırbaş et al.
Scientific reports, 9(1), 19159-19159 (2019-12-18)
From biomarkers to drug carriers, Extracellular Vesicles (EVs) are being used successfully in numerous applications. However, while the subject has been steadily rising in popularity, current methods of isolating EVs are lagging behind, incapable of isolating EVs at a high
Long-circulating poly (ethylene glycol)-modified gelatin nanoparticles for intracellular delivery
Kaul G and Amiji M
Pharmaceutical Research, 19(7), 1061-1067 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service