Skip to Content

94646

Poly(ethylene glycol)

BioUltra, 35,000

greener alternative

Greener Alternative

Synonym(s):

Polyethylene glycol, PEG

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352104
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

BioUltra

form

powder or crystals (or flakes)

mol wt

Mr ~35000

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

insoluble matter:, passes filter test, ≤0.005% peroxides (as H2O2):

pH

5.5-7.0 (25 °C, 50 mg/mL in H2O)

mp

64-66 °C

solubility

H2O: 50 mg/mL at 25 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg, sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤200 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤200 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

λ

50 mg/mL in H2O

UV absorption

λ: 260 nm Amax: 0.12, λ: 280 nm Amax: 0.12

greener alternative category

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

General description

Polyethylene glycol (PEG) is a polyether compound that is used as an additive in cosmetics, pharmaceuticals, and food. It consists of repeated ethylene glycol units[-(CH2CH2O)n].
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. Polyethylene glycol (PEG) is an eco-friendly, biodegradable polymer widely used in pharmaceuticals and cosmetics. Its non-toxic nature and versatility make it a sustainable choice, derived from renewable resources, contributing to greener product formulations. Click here for more information.

Application

Poly(ethylene glycol) has been used:
  • as a component of University of Wisconsin (UW) cold preservation solutions to stabilize the cell membrane of hepatocytes during temperature changes
  • as a component of the supercooling solution to study supercooling preservation technique of the rat liver for transplantation and to protect against hypothermic injury
  • to prepare aqueous phase for the fabrication of multicellular spheroid (MCS) cultures

Biochem/physiol Actions

Polyethylene glycol (PEG) is a synthetic polymer that is used as a gold standard in bioconjugation and nanomedicine to extend blood circulation time and enhance drug efficacy. It acts as an ice-modulator by preventing ice crystallization, which is beneficial for cellular membranes. PEG provides protection against hypothermic injury, like lipid peroxidation and cell membrane injury. PEG-conjugated drugs are considered safe for use in humans and are approved by the U.S. Food and Drug Administration (FDA). PEG has several biomedical applications, including biosensing, imaging, and tissue engineering. It also repels protein adsorption and lowers the liver uptake, which helps in coating nanoparticles.


Still not finding the right product?

Explore all of our products under Poly(ethylene glycol)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library